Skip to Content
Merck
All Photos(1)

Documents

73532

Supelco

2-Methoxyestrone

analytical standard

Synonym(s):

2-MeO-E1, 3-Hydroxy-2-methoxyestra-1,3,5(10)-trien-17-one

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H24O3
CAS Number:
Molecular Weight:
300.39
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:

grade

analytical standard

Quality Level

Assay

≥97.0% (HPLC)

optical activity

[α]/D +182.5±7.5°, c = 0.5 in methanol

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

pharmaceutical

format

neat

storage temp.

2-8°C

SMILES string

C[C@@]12[C@](CCC2=O)([H])[C@]3([H])CCC4=CC(O)=C(OC)C=C4[C@@]3([H])CC1

InChI

1S/C19H24O3/c1-19-8-7-12-13(15(19)5-6-18(19)21)4-3-11-9-16(20)17(22-2)10-14(11)12/h9-10,12-13,15,20H,3-8H2,1-2H3/t12-,13+,15-,19-/m0/s1

InChI key

WHEUWNKSCXYKBU-QPWUGHHJSA-N

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Pictograms

Health hazardEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hannah Oh et al.
British journal of cancer, 117(7), 1070-1078 (2017-08-18)
Prolonged sitting and lower levels of physical activity have been associated with increased levels of parent oestrogens (oestrone and oestradiol), the key hormones in female cancers, in postmenopausal women. However, it is unknown whether sitting and physical activity are associated
Shiying Xu et al.
Ecotoxicology and environmental safety, 209, 111754-111754 (2020-12-16)
There are many kinds of estrogens, and endogenous estrogens produce a variety of estrogen metabolites with similar structure but with different physiological effects after metabolism in vivo. Studies have shown that estrone (E1) widely occurs in the environment and animal-derived
Daniela Pemp et al.
Archives of toxicology, 93(7), 1979-1992 (2019-05-24)
Despite being widely used to investigate 17β-estradiol (E2)-induced mammary gland (MG) carcinogenesis and prevention thereof, estrogen homeostasis and its significance in the female August Copenhagen Irish (ACI) rat model is unknown. Thus, levels of 12 estrogens including metabolites and conjugates
Frank Z Stanczyk et al.
Clinica chimica acta; international journal of clinical chemistry, 484, 223-225 (2018-06-01)
Higher circulating estradiol levels are generally obtained using conventional radioimmunoassays (RIA) compared to liquid chromatography/tandem mass spectrometry (LC-MS/MS) assays, and this has been attributed to the presence of estradiol metabolites that cross-react with the antibody used in the RIA. This
Gieira S Jones et al.
PloS one, 14(5), e0216114-e0216114 (2019-05-09)
Breast density, as estimated by mammography, is a strong risk factor for breast cancer in pre- and postmenopausal women, but the determinants of breast density have not yet been established. The aim of this study was to assess if urinary

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service