Skip to Content
Merck
All Photos(3)

Key Documents

73478

Sigma-Aldrich

Nitromethane

puriss., absolute, over molecular sieve, ≥98.5% (GC)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
CH3NO2
CAS Number:
Molecular Weight:
61.04
Beilstein:
1698205
EC Number:
MDL number:
UNSPSC Code:
12352102
PubChem Substance ID:
NACRES:
NA.21
technique(s):
GC/GC: suitable
bp:
101.2 °C (lit.)
vapor pressure:
2.7 mmHg

vapor density

2.1 (vs air)

vapor pressure

2.7 mmHg

grade

absolute
puriss.

Assay

≥98.5% (GC)

form

liquid

autoignition temp.

784 °F

quality

over molecular sieve

expl. lim.

7.3 %, 33 °F

technique(s)

GC/GC: suitable

impurities

≤0.01% water
water

refractive index

n20/D 1.382 (lit.)
n20/D 1.382

pH

6.4 (20 °C, 0.01 g/L)

bp

101.2 °C (lit.)

mp

−29 °C (lit.)

density

1.127 g/mL at 25 °C (lit.)

SMILES string

C[N+]([O-])=O

InChI

1S/CH3NO2/c1-2(3)4/h1H3

InChI key

LYGJENNIWJXYER-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Nitromethane is an aliphatic nitro compound. It undergoes Michael addition reaction with chalcones in the presence of cinchona alkaloid-derived chiral bifunctional thiourea organocatalyst. Its conversion to N2 in the presence of Co-ZSM5 (Zeolite Socony Mobil-5) has been studied.

Application

Nitromethane may be employed in the preparation of cobalt cage complexes.

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Carc. 2 - Flam. Liq. 3 - Repr. 2

Storage Class Code

4.1A - Other explosive hazardous materials

WGK

WGK 2

Flash Point(F)

95.0 °F - closed cup

Flash Point(C)

35 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

The catalytic chemistry of nitromethane over Co-ZSM5 and other catalysts in connection with the methane-NOxSCR reaction.
Cowan AD, et al.
J. Catal., 176(2), 329-343 (1998)
Loay Awad
Experimental and therapeutic medicine, 19(3), 1835-1843 (2020-02-28)
Chemotherapy and radiation are unable to eliminate all cancer cells, particularly apoptosis-resistant cancer cells, despite their ability to kill cancer cluster cells. Thus, it is important to identify methods that eliminate all cancer cells in order to prevent relapse. Salinomycin
Metal ion encapsulation: cobalt cages derived from polyamines, formaldehyde, and nitromethane.
Geue RJ, et al.
Journal of the American Chemical Society, 106(19), 5478-5488 (1984)
Wenguo Yang et al.
Chemistry, an Asian journal, 7(4), 771-777 (2012-02-10)
Through the cleavage of the C-C bond, the first catalytic tandem conjugate addition-elimination reaction of Morita-Baylis-Hillman C adducts has been presented. Various S(N)2'-like C-, S-, and P-allylic compounds could be obtained with exclusive E configuration in good to excellent yields.
Eagleson M.
Concise Encyclopedia Chemistry, 696-696 (1994)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service