Skip to Content
Merck
All Photos(2)

Key Documents

73400BC

Millipore

Assurance® GDS Adaptor Box

For use with Assurance® GDS Rotor-Gene® Q

Sign Into View Organizational & Contract Pricing

Select a Size

1 EACH
$1,910.00

$1,910.00


Check Cart for Availability

Request more information

Select a Size

Change View
1 EACH
$1,910.00

About This Item

UNSPSC Code:
41121521
NACRES:
NA.74

$1,910.00


Check Cart for Availability

Request more information

Quality Level

product line

Assurance GDS®

technique(s)

PCR: suitable

application(s)

food and beverages

storage temp.

room temp

General description

The Assurance® GDS Adaptor Box is a hardware accessory for use with the Assurance® GDS Rotor-Gene® Q and laptop. As part of the GDS system, the Adaptor Box improves laptop connectivity and PCR run flexibility, allowing one laptop to control up to five Rotor-Genes at one time.

Application

The Assurance GDS® Adaptor Box is an accessory used with the Assurance GDS® system for the detection of pathogenic organisms in foods, ingredients, and environmental samples.

Components

  • Powder Cords for US, UK, EU, Switz, AUS/NZ
  • Ethernet Cable (Cat5/Cat6)
  • Power Supply

Legal Information

Assurance is a registered trademark of Merck KGaA, Darmstadt, Germany
Assurance GDS is a registered trademark of BioControl Systems, Inc.
Rotor-Gene is a registered trademark of Qiagen GmbH

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Assurance® GDS offers integrated solution for fast and accurate food pathogen detection in microbiology testing.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service