Skip to Content
Merck
All Photos(1)

Key Documents

73297

Sigma-Aldrich

Stearic acid 50

tested according to Ph. Eur.

Synonym(s):

Palmitic acid – stearic acid mixture, Acidum stearicum 50, Fatty acids C16-18, Stearic acid - palmitic acid mixture

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Beilstein:
608585
EC Number:
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.25
Product 73297 is not currently sold in your country. Contact Technical Service

biological source

palm oil

Quality Level

Agency

USP/NF
tested according to Ph. Eur.

Assay

≥90% (sum of steric acid and palmitic acid)

form

liquid

functional group

carboxylic acid

shipped in

ambient

storage temp.

room temp

SMILES string

CCCCCCCCCCCCCCCCCC(O)=O

InChI

1S/C18H36O2/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18(19)20/h2-17H2,1H3,(H,19,20)

InChI key

QIQXTHQIDYTFRH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Other Notes

mixture of fatty acids, consisting mainly of stearic acid and 40-60% palmitic acid (acc. to Ph.Eur.)

Storage Class Code

13 - Non Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Palle J Pedersen et al.
Journal of medicinal chemistry, 53(9), 3782-3792 (2010-04-22)
The design of retinoid phospholipid prodrugs is described based on molecular dynamics simulations and cytotoxicity studies of synthetic retinoid esters. The prodrugs are degradable by secretory phospholipase A(2) IIA and have potential in liposomal drug delivery targeting tumors. We have
Jih-Jung Chen et al.
Journal of natural products, 72(2), 223-228 (2009-02-06)
Six new compounds, including five new seco-abietane diterpenoids, 12-deoxy-seco-hinokiol methyl ester (1), 12-deoxy-11,12-dihydro-seco-hinokiol methyl ester (2), callicarpic acid A (3), 9alpha-hydroxycallicarpic acid A (4), and callicarpic acid B (5), and a new phenylethanoid derivative, 4-hydroxyphenethyl tetradecanoate (6), have been isolated
Bhadreshkumar B Maisuria et al.
Bioorganic & medicinal chemistry, 19(9), 2918-2926 (2011-04-16)
Homologous dicarboxyl dendritic amphiphiles-RCONHC(CH(3))(CH(2)CH(2)COOH)(2), 4(n); and ROCONHC(CH(3))(CH(2)CH(2)COOH)(2), 5(n), where R=n-C(n)H(2)(n)(+1) and n=13-22 carbon atoms-were synthesized. Critical micelle concentrations (CMCs) in aqueous triethanolamine solutions and at pH 7.4 were measured along with hemolytic activity (effective concentrations, EC(10)) in phosphate-buffered saline (PBS).
Arpita Chatterjee et al.
Antimicrobial agents and chemotherapy, 52(1), 220-224 (2007-10-24)
Cholera toxin (CT) is an archetypal bacterial toxin that binds with a high affinity to the receptor ganglioside GM1 on the intestinal epithelial surface and that causes the severe watery diarrhea characteristic of the disease cholera. Blockage of the interaction
Marcy J Balunas et al.
Journal of natural products, 69(4), 700-703 (2006-04-29)
Natural product drug discovery efforts frequently utilize noncellular screening assays. Fatty acids are commonly found in natural product extracts, and some have been shown to interfere with noncellular assays. Several pure fatty acids were tested using a noncellular aromatase assay

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service