Skip to Content
Merck
All Photos(1)

Documents

73100

Sigma-Aldrich

2-Nitrobenzoyl chloride

purum, ≥97.0% (T)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
O2NC6H4COCl
CAS Number:
Molecular Weight:
185.56
Beilstein:
777991
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

grade

purum

Assay

≥97.0% (T)

form

liquid

refractive index

n20/D 1.569 (lit.)
n20/D 1.569

bp

148-149 °C/9 mmHg (lit.)

mp

17-20 °C (lit.)

density

1.404 g/mL at 25 °C (lit.)

SMILES string

[O-][N+](=O)c1ccccc1C(Cl)=O

InChI

1S/C7H4ClNO3/c8-7(10)5-3-1-2-4-6(5)9(11)12/h1-4H

InChI key

BWWHTIHDQBHTHP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service