Skip to Content
Merck
All Photos(1)

Documents

72777

Supelco

trans-Quercus lactone

analytical standard

Synonym(s):

trans-5-Butyl-4-methyloxolan-2-one, trans-5-Butyldihydro-4-methyl-2(3H)-furanone, trans-Methyloctanolide, trans-Oak lactone, trans-Whiskey lactone, trans-beta-Methyl-γ-octalactone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H16O2
CAS Number:
Molecular Weight:
156.22
Beilstein:
4661547
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥94.0% (GC)

shelf life

limited shelf life, expiry date on the label

application(s)

cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care

format

neat

SMILES string

CCCC[C@@H]([C@H](C)C1)OC1=O

InChI

1S/C9H16O2/c1-3-4-5-8-7(2)6-9(10)11-8/h7-8H,3-6H2,1-2H3/t7-,8+/m1/s1

InChI key

WNVCMFHPRIBNCW-SFYZADRCSA-N

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service