Skip to Content
Merck
All Photos(1)

Key Documents

72659

Millipore

Anthracis Selective Supplement

suitable for microbiology

Sign Into View Organizational & Contract Pricing

Select a Size

5 AMP
$135.00

$135.00


Estimated to ship on24 May 2025


Request more information

Select a Size

Change View
5 AMP
$135.00

About This Item

UNSPSC Code:
41171614
NACRES:
NA.85

$135.00


Estimated to ship on24 May 2025


Request more information

sterility

sterile

Quality Level

form

powder

shelf life

limited shelf life, expiry date on the label

application(s)

clinical testing
environmental
food and beverages
veterinary

microbiology

storage temp.

2-8°C

suitability

Bacillus anthracis

General description

Polymixin is an inhibitory agent allowing growth of Bacillus anthracis while inhibiting contaminants. Lysozyme specifically suppressses the growth of Gram negative contaminants.

Application

inhibitory agent allowing growth of Bacillus anthracis while inhibiting contaminants like Gram negatives

Components

(per vial sufficient for 1000 mL medium)
Lysozyme 300000 units, Polymyxin B sulfate 30000 units

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Resp. Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service