Skip to Content
Merck
All Photos(1)

Key Documents

71895

Sigma-Aldrich

4-Hydrazino-7-nitro-benzofurazan hydrazine adduct

for fluorescence, ≥97.0% (HPLC)

Synonym(s):

7-Nitro-4-benzofurazanyl-hydrazine hydrazine adduct, NBD-H

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H5N5O3 · NH2NH2
CAS Number:
Molecular Weight:
227.18
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

grade

for fluorescence

Assay

≥97.0% (HPLC)

fluorescence

λex 445 nm; λem 525 nm in chloroform (after derivatization with formaldehyde)

storage temp.

2-8°C

SMILES string

NN.NNc1ccc([N+]([O-])=O)c2nonc12

InChI

1S/C6H5N5O3.H4N2/c7-8-3-1-2-4(11(12)13)6-5(3)9-14-10-6;1-2/h1-2,8H,7H2;1-2H2

InChI key

BLIOAKMSUDUCKB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Other Notes

Fluorogenic derivatizing reagent for aldehydes ketones

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Carc. 1B - Muta. 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S. Uzu et al.
Analyst, 115, 1477-1477 (1990)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service