Skip to Content
Merck
All Photos(1)

Key Documents

71809

Supelco

Poly(hexafluoropropylene oxide)

suitable for mass spectrometry (MS)

Synonym(s):

Poly(perfluoropropylene oxide)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HO[CF(CF3)CF2O]nH
CAS Number:
MDL number:
UNSPSC Code:
12000000

vapor pressure

4 × 10-7 ( 20 °C)

technique(s)

mass spectrometry (MS): suitable

refractive index

n20/D 1.297

SMILES string

OC(F)(F)C(F)(F)F.OC(F)(F)C(O)(F)C(F)(F)F

Looking for similar products? Visit Product Comparison Guide


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

B Uzan et al.
Journal of biochemical and biophysical methods, 35(1), 49-55 (1997-08-01)
O2 can be gently removed from solutions by contact with a "bulk fluid membrane" of the viscous and nearly inert perfluoro polyether Fomblin Y. A volume of Fomblin dissolves approximately 20 times more O2 than an equal volume of water.
Adriana M Vieira Silveira et al.
Brazilian dental journal, 18(4), 299-304 (2008-02-19)
The number of appointments necessary to treat infected root canals is one of the most controversial issues in endodontics. This study evaluated, in dogs, the response of the periradicular tissues to the endodontic treatment of infected root canals performed in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service