70135
DCLS Agar
suitable for microbiology, NutriSelect® Plus
Synonym(s):
Deoxycholate Citrate Lactose Sucrose Agar
About This Item
Recommended Products
form
powder
Quality Level
shelf life
limited shelf life, expiry date on the label
composition
agar, 12.0 g/L
lactose, 5.0 g/L
neutral red, 0.03 g/L
sodium citrate, 10.5 g/L
sodium desoxycholate, 2.5 g/L
sodium thiosulfate, 5.0 g/L
special peptone, 10.0 g/L
sucrose, 5.0 g/L
manufacturer/tradename
NutriSelect® Plus
technique(s)
microbiological culture: suitable
final pH
7.2±0.2 (25 °C)
application(s)
clinical testing
environmental
food and beverages
microbiology
suitability
selective and differential for Salmonella spp.
selective and differential for Shigella spp.
selective and differential for Vibrio spp.
selective and differential for Yersinia spp.
Application
Preparation Note
Footnote
The designations basic, plus, or prime are added to indicate the quality control level, from basic quality control to standard QC plus to prime for full regulatory compliance.
Legal Information
Storage Class Code
13 - Non Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Articles
Vibrios are motile, curved or comma-shaped bacilli and have a single polar flagella with sheet proteins. They are often found in open water, freshwater and saltwater.
Salmonella, with 2,300+ serotypes, causes half of food-borne illnesses, often from dairy, poultry, and eggs.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service