Skip to Content
Merck
All Photos(1)

Key Documents

69499

Supelco

3-Propylmalic acid

≥95.0% (HPLC)

Synonym(s):

β-Propylmalic acid, (±)-2-Hydroxy-3-propylbutandioic acid, 2-Hydroxy-3-propylsuccinic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H12O5
CAS Number:
Molecular Weight:
176.17
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

Quality Level

Assay

≥95.0% (HPLC)

application(s)

clinical testing

format

neat

storage temp.

2-8°C

SMILES string

OC(C(CCC)C(O)C(O)=O)=O

InChI

1S/C7H12O5/c1-2-3-4(6(9)10)5(8)7(11)12/h4-5,8H,2-3H2,1H3,(H,9,10)(H,11,12)

InChI key

LOLHYFQEDPGSHZ-UHFFFAOYSA-N

Biochem/physiol Actions

Metabolite of the glyoxylate and dicarboxylate metabolism.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

N-PROPYLMALATE SYNTHETASE.
K IMAI et al.
The Journal of biological chemistry, 238, 3193-3198 (1963-10-01)
Synthesis and breakdown of n-propylmalic acid.
Imai, K., et al.
Biochimica et Biophysica Acta, 81, 367-376 (1964)
J R Stern et al.
Journal of bacteriology, 98(1), 147-151 (1969-04-01)
A mutant strain of Salmonella typhimurium (SL 1634 dml-51) capable of growth on d-malate as sole carbon source was shown to produce d-malic enzyme. This enzyme was absent in the parent wild-type strain which was unable to grow on d-malate.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service