Skip to Content
Merck
All Photos(1)

Key Documents

68740

Sigma-Aldrich

3-Methyl-1-phenyl-2-pyrazoline-5-one

purum, ≥98.0% (NT)

Synonym(s):

5-Methyl-2-phenyl-2,4-dihydro-3H-pyrazol-3-one

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H10N2O
CAS Number:
Molecular Weight:
174.20
Beilstein:
609575
EC Number:
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:

grade

purum

Assay

≥98.0% (NT)

bp

287 °C/265 mmHg (lit.)

mp

126-128 °C (lit.)
126-128 °C

SMILES string

CC1=NN(C(=O)C1)c2ccccc2

InChI

1S/C10H10N2O/c1-8-7-10(13)12(11-8)9-5-3-2-4-6-9/h2-6H,7H2,1H3

InChI key

QELUYTUMUWHWMC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

replaced by

Product No.
Description
Pricing

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hidehiko Nakagawa et al.
Bioorganic & medicinal chemistry letters, 16(23), 5939-5942 (2006-09-26)
We synthesized various 3-methyl-1-phenyl-5-pyrazolone (edaravone) derivatives and evaluated their oxidation potential and hydroxyl radical scavenging activity. It was found 3-methyl-1-(pyridin-2-yl)-5-pyrazolone had a much higher ability to scavenge the radical than did edaravone itself. Its efficient radical scavenging activity was assumed
Xin-Gang Du et al.
Journal of natural products, 73(8), 1422-1426 (2010-08-04)
Two new dimeric xanthone O-glycosides, puniceasides A (1) and B (2), a new trimeric O-glycoside, puniceaside C (3), and two new trimeric C-glycosides, puniceasides D (4) and E (5), together with 12 known xanthones were isolated from the entire plant
Ajmal Ahmad et al.
Molecular and cellular biochemistry, 367(1-2), 215-225 (2012-06-01)
Stroke is a life-threatening disease with major cause of mortality and morbidity worldwide. The neuronal damage following cerebral ischemia is a serious risk to stroke patients. Oxidative stress and apoptotic damage play an important role in cerebral ischemic pathogenesis and
Xian-Wen Yang et al.
Journal of natural products, 70(4), 521-525 (2007-03-09)
Six new mixed lignan-neolignans and 20 known compounds were isolated from the whole plant of Tarenna attenuata. By analysis of physical and spectroscopic data, the structures of the new compounds were elucidated as (1R,5R,6R)-6-{4-O-[2-(1-(4-hydroxy-3-methoxyphenyl))glycerol]-3,5-dimethoxyphenyl}-3,7-dioxabicyclo[3.3.0]octan-2-one (1), 5' '-methoxyhedyotisol A (2), 4'
Konstantin Chegaev et al.
Journal of medicinal chemistry, 52(2), 574-578 (2008-12-31)
A new series of polyvalent drugs obtained by joining edaravone with NO-donor moieties is described. All compounds display high antioxidant power together with NO-dependent vasodilator properties. The analysis of a number of molecular descriptors shows that the antioxidant activity, which

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service