Skip to Content
Merck
All Photos(2)

Key Documents

68723

Supelco

Triazamate

PESTANAL®, analytical standard

Synonym(s):

Ethyl (3-tert-butyl-1-dimethylcarbamoyl-1H-1,2,4-triazol-5-ylthio)acetate, Ethyl 2-{{1-[(dimethylamino)carbonyl]-3-(1,1-dimethylethyl)-1H-1,2,4-triazol-5-yl}thio}acetate

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
$602.00

$602.00


Estimated to ship on29 May 2025


Request a Bulk Order

Select a Size

Change View
100 MG
$602.00

About This Item

Empirical Formula (Hill Notation):
C13H22N4O3S
CAS Number:
Molecular Weight:
314.40
Beilstein:
8422595
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

$602.00


Estimated to ship on29 May 2025


Request a Bulk Order

grade

analytical standard

Quality Level

product line

PESTANAL®

Assay

≥98.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

composition

carbon content, 48.66-50.65%
nitrogen content, 17.46-18.18%

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

mp

52-56 °C

application(s)

agriculture
environmental

format

neat

SMILES string

CCOC(=O)CSc1nc(nn1C(=O)N(C)C)C(C)(C)C

InChI

1S/C13H22N4O3S/c1-7-20-9(18)8-21-11-14-10(13(2,3)4)15-17(11)12(19)16(5)6/h7-8H2,1-6H3

InChI key

NKNFWVNSBIXGLL-UHFFFAOYSA-N

General description

Triazamate is a carbamoyl triazole compound, widely used as an insecticide and its mode of action involves the inhibition of cholinesterase in insect pests.[1]

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.
Triazamate may be used as an analytical reference standard for the quantification of the analyte in water samples using gas chromatography technique.[2]

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Skull and crossbonesEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Inhalation - Acute Tox. 3 Oral - Aquatic Acute 1 - Eye Irrit. 2

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

372.2 °F

Flash Point(C)

189 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Aphids as Crop Pests (2007)
Development and validation of a multiresidue method for determination of 82 pesticides in water using GC
Mamun R.IM, et al.
Journal of Separation Science, 32(4), 559-574 (2009)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service