Skip to Content
Merck
All Photos(1)

Key Documents

68678

Supelco

6-Chloropyridine-3-carboxylic acid

PESTANAL®, analytical standard

Synonym(s):

6-Chloronicotinic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H4ClNO2
CAS Number:
Molecular Weight:
157.55
Beilstein:
115993
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

Assay

≥98.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

mp

185-190 °C
190 °C (dec.) (lit.)

application(s)

agriculture
environmental

format

neat

SMILES string

OC(=O)c1ccc(Cl)nc1

InChI

1S/C6H4ClNO2/c7-5-2-1-4(3-8-5)6(9)10/h1-3H,(H,9,10)

InChI key

UAWMVMPAYRWUFX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

6-Chloropyridine-3-carboxylic acid is a metabolite of the neonicotinoid pesticides, imidacloprid and acetamiprid and occurs in different environmental matrices.(3) It is widely used in agricultural crop protection, domestic insect control and environmental pest management.[1]
Find more information here: Neonicotinoids

Application

6-Chloropyridine-3-carboxylic acid may be used as an analytical reference standard for the quantification of the analyte in biological samples[1] and animal derived foods[2] using different chromatography techniques.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Qualitative profiling and quantification of neonicotinoid metabolites in human urine by liquid chromatography coupled with mass spectrometry
Taira K, et al.
Testing, 8(11) (2013)
Qualitative profiling and quantification of neonicotinoid metabolites in human urine by liquid chromatography coupled with mass spectrometry
Taira K, et al.
PLoS ONE, 8(11) (2013)
A green HPLC technique with a 100\% water mobile phase for detecting imidacloprid and its metabolite 6-chloronicotinic acid
Furusawa N
Current Chemistry Letters, 5(1), 27-32 (2016)
Gretty Ettiene et al.
Electrophoresis, 33(19-20), 2969-2977 (2012-09-22)
A sensitive and reliable method based on MEKC has been developed and validated for trace determination of neonicotinoid insecticides (thiamethoxam, acetamiprid, and imidacloprid) and the metabolite 6-chloronicotinic acid in water and soil matrices. Optimum separation of the neonicotinoid insecticides was
Romina Zabar et al.
Chemosphere, 85(5), 861-868 (2011-08-02)
This work describes for the first time the photolytic and photocatalytic degradation of 6-chloronicotinic acid (6CNA) in double deionised water, which is a degradation product of neonicotinoid insecticides imidacloprid and acetamiprid, and it is known to appear in different environmental

Protocols

Learn more about Neonicotinoids - active substances used in plant protection products to control harmful insects.

Analysis of banned neonicotinoid insecticides from dandelion blossoms using QuEChERS and LC-MS.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service