Skip to Content
Merck
All Photos(1)

Key Documents

67862

Supelco

Homogentisic acid

analytical standard

Synonym(s):

2,5-Dihydroxyphenylacetic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(HO)2C6H3CH2CO2H
CAS Number:
Molecular Weight:
168.15
Beilstein:
2692860
EC Number:
MDL number:
UNSPSC Code:
41116107

grade

analytical standard

Assay

≥98.0% (GC)

shelf life

limited shelf life, expiry date on the label

mp

150-152 °C (lit.)

application(s)

clinical testing

format

neat

storage temp.

2-8°C

SMILES string

OC(=O)Cc1cc(O)ccc1O

InChI

1S/C8H8O4/c9-6-1-2-7(10)5(3-6)4-8(11)12/h1-3,9-10H,4H2,(H,11,12)

InChI key

IGMNYECMUMZDDF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Homogentisic acid is a marker of the botanic origin of strawberry tree honey.

Application

Homogentisic acid may be used as an analytical reference standard for the quantification of the analyte in the following:
  • Urine matrices using reversed-phase liquid chromatography tandem mass spectrometry (LC-MS/MS).
  • Strawberry tree honey samples using reversed phase-high performance liquid chromatography (RP-HPLC) and ion chromatography (IC).

It may also be used as an internal standard for the determination of ascorbic acid (AA) and uric acid (UA) in human plasma samples using high-pressure liquid chromatography–electrochemical detection (HPLC–ECD).

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Fast HPLC?ECD analysis of ascorbic acid, dehydroascorbic acid and uric acid
Li X, et al.
Journal of Chromatography. B, Biomedical Sciences and Applications, 877(10), 853-856 (2009)
Direct chromatographic methods for the rapid determination of homogentisic acid in strawberry tree (Arbutus unedo L.) honey
Scanu R, et al.
Journal of Chromatography A, 1090(1-2), 76-80 (2005)
Urine homogentisic acid and tyrosine: simultaneous analysis by liquid chromatography tandem mass spectrometry
Hughes TA, et al.
Journal of Chromatography. B, Biomedical Sciences and Applications, 963, 106-112 (2014)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service