67712
O-Methylisourea hemisulfate salt
≥99.0% (T)
Synonym(s):
OMI®
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Assay
≥99.0% (T)
mp
163-167 °C (lit.)
164-167 °C
solubility
H2O: 0.1 g/mL, clear
SMILES string
COC(N)=N.COC(N)=N.OS(O)(=O)=O
InChI
1S/2C2H6N2O.H2O4S/c2*1-5-2(3)4;1-5(2,3)4/h2*1H3,(H3,3,4);(H2,1,2,3,4)
InChI key
QSCPQKVWSNUJLJ-UHFFFAOYSA-N
Gene Information
human ... OPRD1(4985) , OPRK1(4986) , OPRM1(4988)
rat ... Oprd1(24613) , Oprm1(25601)
Looking for similar products? Visit Product Comparison Guide
Legal Information
OMI is a registered trademark of Merck KGaA, Darmstadt, Germany
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service