Skip to Content
Merck
All Photos(3)

Key Documents

67000BC

Millipore

TRANSIA® AG Listeria Kit

suitable for ELISA testing in Environmental, and Food and Beverages

Sign Into View Organizational & Contract Pricing

Select a Size

96 TESTS
$1,480.00
504 TESTS
$3,800.00

$1,480.00


Check Cart for Availability

Request a Bulk OrderRequest more information

Select a Size

Change View
96 TESTS
$1,480.00
504 TESTS
$3,800.00

About This Item

UNSPSC Code:
41116129
NACRES:
NA.85

$1,480.00


Check Cart for Availability

Request a Bulk OrderRequest more information

Product Name

TRANSIA® AG Listeria, BioControl, ELISA test for detection of Listeria in food and environmental samples

Agency

AOAC® (OMA)
AOAC® (PTM)

product line

TRANSIA®

form

liquid

shelf life

6‑15 mo.

packaging

pkg of 504
pkg of 96

manufacturer/tradename

BioControl Systems 67000-504
BioControl Systems 67000-96
TRANSIA®

technique(s)

ELISA: suitable

input

food(s)

application(s)

food and beverages
pathogen testing

storage temp.

2-8°C

suitability

Listeria spp.

General description

TRANSIA AG Listeria is an enzyme immunoassay that detects Listeria in environmental samples and a variety of foods including fish, fruit and fruit products, meat and meat products (red meats, pork, and poultry), pasta, dairy products, eggs and egg products. The test is intended for use by laboratory personnel with appropriate microbiology training.

Application

AOAC Official Method 996.14, AOAC Performance Tested Method 060802

Features and Benefits

  • Labor saving single-step enrichment in Demi-Fraser Broth for all samples.
  • Validated to provide results in 48 hours for both environmental surfaces and food samples.
  • Breakaway wells allow you to use only the number of tests required for your samples, helping to reduce waste and keep testing costs low.
  • Available in convenient 96 test or 504 test kits.

Components

Test wells, wash solution concentrate, substrate, conjugate, positive control, stop solution

Testing Method

ELISA

Legal Information

AOAC is a registered trademark of AOAC International
TRANSIA is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Aquatic Chronic 3 - Eye Dam. 1 - Repr. 2 - Skin Irrit. 2 - Skin Sens. 1

Storage Class Code

10 - Combustible liquids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Related Content

TRANSIA AG (Enzyme ImmunoAssay) and TRANSIA® PLATE incorporate proven antibody-antigen "sandwich" assay technology for the detection of food borne pathogens in both product and environmental samples

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service