Skip to Content
Merck
All Photos(1)

Key Documents

66030

Supelco

2-Methyl-1-butene

analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH2C(CH3)=CH2
CAS Number:
Molecular Weight:
70.13
Beilstein:
505975
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

vapor pressure

9.98 psi ( 20 °C)

Assay

≥99.5% (GC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

refractive index

n20/D 1.378 (lit.)
n20/D 1.378

bp

31 °C (lit.)

mp

−137 °C (lit.)

density

0.65 g/mL at 25 °C (lit.)

application(s)

environmental
petroleum

format

neat

storage temp.

2-8°C

SMILES string

CCC(C)=C

InChI

1S/C5H10/c1-4-5(2)3/h2,4H2,1,3H3

InChI key

MHNNAWXXUZQSNM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

2-Methyl-1-butene, an isoamylene grouped under the class of volatile organic compounds (VOCs), is typically a heavy, active component of C5 olefin fractions of gasoline.

Application

2-Methyl-1-butene may be used as an analytical standard for the determination of the analyte in poly(methyl methacrylate) kitchen utensils, household waste and building materials, products of vapor phase olefin-ozone reactions, and Fischer–Tropsch synthesis products by various chromatography techniques.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Pictograms

Flame

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Liq. 1

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

-34.6 °F - closed cup

Flash Point(C)

-37 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Isobaric Vapor- Liquid Equilibria of Binary and Ternary Mixtures of Heavy Ethers, Isoamylenes, and Alcohols. 2. Tertiary Amyl Ethyl Ether, 2-Methyl-1-butene, and Ethanol
Everson RC and Jansen W
Journal of Chemical and Engineering Data, 46(2), 247-250 (2001)
Chromatographic-mass spectrometric analysis of Fischer-Tropsch synthesis products
Zaikin VG and Borisov RS
Journal of Analytical Chemistry, 57(6), 544-551 (2002)
Effect of macroreticular acidic ion-exchange resins on 2-methyl-1-butene and 2-methyl-2-butene mixture oligomerization
Granollers M, et al.
Applied Catalysis A: General, 435, 163-171 (2012)
A STUDY OF THE PRODUCTS OF THE REACTIONS OF OZONE WITH OLEFINS IN THE VAPOR PHASE AS DETERMINED BY GAS-LIQUID CHROMATOGRAPHY
Vrbaski T and Cvetanovic RJ
Canadian Journal of Chemistry, 38(7), 1063-1069 (1960)
Identification of volatile (micro) biological compounds from household waste and building materials by thermal desorption-capillary gas chromatography-mass spectroscopy
Wilkins CK and Larsen K
Journal of High Resolution Chromatography, 18(6), 373-377 (1995)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service