Skip to Content
Merck
All Photos(2)

Key Documents

65250

Sigma-Aldrich

Dimethyl 4-methoxyphthalate

≥97.0% (GC)

Synonym(s):

Dimethyl 4-methoxybenzene-1,2-dicarboxylate

Sign Into View Organizational & Contract Pricing

Select a Size

10 ML
$514.00

$514.00


Estimated to ship on26 March 2025


Request a Bulk Order

Select a Size

Change View
10 ML
$514.00

About This Item

Empirical Formula (Hill Notation):
C11H12O5
CAS Number:
Molecular Weight:
224.21
Beilstein:
2373838
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Assay:
≥97.0% (GC)

$514.00


Estimated to ship on26 March 2025


Request a Bulk Order

Assay

≥97.0% (GC)

refractive index

n20/D 1.531

bp

140 °C/0.1 mmHg (lit.)

density

1.225 g/mL at 20 °C (lit.)

functional group

ester

SMILES string

COC(=O)c1ccc(OC)cc1C(=O)OC

InChI

1S/C11H12O5/c1-14-7-4-5-8(10(12)15-2)9(6-7)11(13)16-3/h4-6H,1-3H3

InChI key

WKSKDXDNBSJCIM-UHFFFAOYSA-N

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

228.2 °F - closed cup

Flash Point(C)

109.00 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service