Skip to Content
Merck
All Photos(1)

Documents

650188

Sigma-Aldrich

Methanol solution

suitable for HPLC, contains Methanol:Dimethyl sulfoxide 1:1 (v/v)

Synonym(s):

Dimethyl sulfoxide:Methanol solution

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
CH4O
CAS Number:
Molecular Weight:
32.04
Beilstein:
3662861
MDL number:
UNSPSC Code:
12190000
PubChem Substance ID:

product name

Methanol solution, (Methanol:Dimethyl sulfoxide 1:1 (v/v))

vapor density

1.11 (vs air)

Quality Level

vapor pressure

410 mmHg

form

liquid

expl. lim.

5.5-44 % (lit.)

concentration

(Methanol:Dimethyl sulfoxide 1:1 (v/v))

technique(s)

HPLC: suitable

bp

65.4 °C (lit.)

mp

-98 °C (lit.)

density

0.954 g/mL at 20 °C

application(s)

food and beverages

format

mixture

SMILES string

CO

InChI

1S/CH4O/c1-2/h2H,1H3

InChI key

OKKJLVBELUTLKV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

The product is a 1:1(v/v) solution of methanol and dimethylsulfoxide that can be employed for HPLC and UHPLC (Ultra High performance Liquid Chromatography) techniques. This pre-blended solvent is of high purity that reduces time-consuming preparation and prevents instrument downtime because of impure mobile phases.

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

71.6 °F - closed cup

Flash Point(C)

22 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kentaro Fujiwara et al.
EJNMMI research, 4, 29-29 (2014-07-10)
ROBO1 is a membrane protein that functions in axon guidance. ROBO1 contributes to tumour metastasis and angiogenesis and may have potential as a target protein of immunotherapy because ROBO1 is specifically expressed at high levels in hepatocellular carcinoma. In this
Haruka Oda et al.
Biomaterials, 56, 86-91 (2015-05-03)
A large number of lineage-committed progenitor cells are required for advanced regenerative medicine based on cell engineering. Due to their ability to differentiate into multiple cells lines, multipotent stem cells have emerged as a vital source for generating transplantable cells
Timo Heidt et al.
Circulation research, 115(2), 284-295 (2014-05-03)
Macrophages populate the steady-state myocardium. Previously, all macrophages were thought to arise from monocytes; however, it emerged that, in several organs, tissue-resident macrophages may self-maintain through local proliferation. Our aim was to study the contribution of monocytes to cardiac-resident macrophages
Mathias Schmelcher et al.
The Journal of antimicrobial chemotherapy, 70(5), 1453-1465 (2015-01-30)
In the light of increasing drug resistance in Staphylococcus aureus, bacteriophage endolysins [peptidoglycan hydrolases (PGHs)] have been suggested as promising antimicrobial agents. The aim of this study was to determine the antimicrobial activity of nine enzymes representing unique homology groups
B Alkhatib et al.
European cells & materials, 28, 98-110 (2014-09-13)
Excessive mechanical loading or acute trauma to intervertebral discs (IVDs) is thought to contribute to degeneration and pain. However, the exact mechanisms by which mechanical injury initiates and promotes degeneration remain unclear. This study investigates biochemical changes and extracellular matrix

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service