Skip to Content
Merck
All Photos(3)

Key Documents

63112

Supelco

Magnesium ionophore VI

Selectophore, function tested

Synonym(s):

1,3,5-Tris[10-(1-adamantyl)-7,9-dioxo-6,10-diazaundecyl]benzene, ETH 5506

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
$1,940.00
100 MG
$14,150.00

$1,940.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
10 MG
$1,940.00
100 MG
$14,150.00

About This Item

Empirical Formula (Hill Notation):
C63H96N6O6
CAS Number:
Molecular Weight:
1033.47
MDL number:
UNSPSC Code:
26111700
PubChem Substance ID:
NACRES:
NB.61

$1,940.00


Check Cart for Availability

Request a Bulk Order

description

for ion-selective electrodes

Quality Level

product line

Selectophore

Assay

≥99.0% (HPLC)

quality

function tested

SMILES string

CN(C(=O)CC(=O)NCCCCCc1cc(CCCCCNC(=O)CC(=O)N(C)C23CC4CC(CC(C4)C2)C3)cc(CCCCCNC(=O)CC(=O)N(C)C56CC7CC(CC(C7)C5)C6)c1)C89CC%10CC(CC(C%10)C8)C9

InChI

1S/C63H96N6O6/c1-67(61-34-46-22-47(35-61)24-48(23-46)36-61)58(73)31-55(70)64-16-10-4-7-13-43-19-44(14-8-5-11-17-65-56(71)32-59(74)68(2)62-37-49-25-50(38-62)27-51(26-49)39-62)21-45(20-43)15-9-6-12-18-66-57(72)33-60(75)69(3)63-40-52-28-53(41-63)30-54(29-52)42-63/h19-21,46-54H,4-18,22-42H2,1-3H3,(H,64,70)(H,65,71)(H,66,72)

InChI key

ODMHCQNFQKBJOO-UHFFFAOYSA-N

General description

Visit our Sensor Applications portal to learn more.

Application

Selected application examples

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Other Notes

Lipophilic magnesium-selective ionophore (log PTLC=8.5) with sufficient selectivity for Mg2+ against the main interfering cations for serum measurments[1][2]

Legal Information

Selectophore is a trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Development of magnesium-selective ionophores
O'Donnell, J., et al.
Analytica Chimica Acta, 281, 129-134 (1993)
Potentiometric microelectrodes as sensors and detectors. Magnesium-selective electrodes as sensors, and Hofmeister electrodes as detectors for histmaine in capillary electrophoresis
Spichiger, U.E., Fakler, A.
Electrochimica Acta, 42, 3137-3145 (1997)
A Comparison of Neutral Mg2+-Selective Ionophores in Solvents Polymeric Membranes: Complex Stoichiometry
Zhang, W., et al.
Analytical Sciences, 16, 11-18 (2000)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service