Skip to Content
Merck
All Photos(1)

Key Documents

62881

Supelco

2-[(RS)-(4-Bromophenyl)phenylmethoxy]-N,N-dimethylethanamine hydrochloride

pharmaceutical impurity standard

Synonym(s):

Diphenhydramine impurity C (PhEur), 2-[(4-Bromophenyl)phenylmethoxy]ethyl(dimethyl)ammonium chloride, Bromazine hydrochloride, Bromodiphenhydramine hydrochloride, NSC 36113

Sign Into View Organizational & Contract Pricing

Select a Size

50 MG
$985.00

$985.00


Estimated to ship on31 May 2025


Request a Bulk Order

Select a Size

Change View
50 MG
$985.00

About This Item

Empirical Formula (Hill Notation):
C17H20BrNO · HCl
CAS Number:
Molecular Weight:
370.71
Beilstein:
3754363
EC Number:
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:
NACRES:
NA.24

$985.00


Estimated to ship on31 May 2025


Request a Bulk Order

Quality Level

Assay

≥97.0% (HPLC)
97.0-100.0% (AT)

shelf life

limited shelf life, expiry date on the label

impurities

≤1.0% water

application(s)

forensics and toxicology
pharmaceutical (small molecule)

format

neat

SMILES string

BrC(C=C1)=CC=C1C(OCCN(C)C)C2=CC=CC=C2.Cl

InChI

1S/C17H20BrNO.ClH/c1-19(2)12-13-20-17(14-6-4-3-5-7-14)15-8-10-16(18)11-9-15;/h3-11,17H,12-13H2,1-2H3;1H

InChI key

ZQDJSWUEGOYDGT-UHFFFAOYSA-N

Gene Information

human ... HRH1(3269)

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service