Skip to Content
Merck
All Photos(1)

Key Documents

61224

Supelco

N-Succinimidyl 4-(dimethylamino)benzoate

for HPLC derivatization, LiChropur, ≥98.0% (HPLC)

Synonym(s):

1-{[4-(Dimethylamino)benzoyl]oxy}pyrrolidine-2,5-dione, 4-(Dimethylamino)benzoic acid 2,5-dioxo-1-pyrrolidinyl ester, 4-(Dimethylamino)benzoic acid N-hydroxysuccinimide ester

Sign Into View Organizational & Contract Pricing

Select a Size

500 MG
$1,060.00

$1,060.00


Estimated to ship on01 June 2025


Request a Bulk Order

Select a Size

Change View
500 MG
$1,060.00

About This Item

Empirical Formula (Hill Notation):
C13H14N2O4
CAS Number:
Molecular Weight:
262.26
Beilstein:
1479401
MDL number:
UNSPSC Code:
12000000
PubChem Substance ID:
NACRES:
NA.22

$1,060.00


Estimated to ship on01 June 2025


Request a Bulk Order

grade

derivatization grade ((HPLC))
for HPLC derivatization

Quality Level

Assay

≥98.0% (HPLC)

form

solid

quality

LiChropur

technique(s)

HPLC: suitable

SMILES string

CN(C)C1=CC=C(C(ON(C(CC2)=O)C2=O)=O)C=C1

InChI

1S/C13H14N2O4/c1-14(2)10-5-3-9(4-6-10)13(18)19-15-11(16)7-8-12(15)17/h3-6H,7-8H2,1-2H3

InChI key

FNGFZOAQGYOQTG-UHFFFAOYSA-N

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

LiChropur is a trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service