Skip to Content
Merck
All Photos(2)

Key Documents

61193

Supelco

Copper(II) ionophore I

Selectophore

Synonym(s):

o-XBDiBDTC, o-Xylylenebis(N,N-diisobutyldithiocarbamate)

Sign Into View Organizational & Contract Pricing

Select a Size

50 MG
$523.00

$523.00


Estimated to ship on15 April 2025


Request a Bulk Order

Select a Size

Change View
50 MG
$523.00

About This Item

Empirical Formula (Hill Notation):
C26H44N2S4
CAS Number:
Molecular Weight:
512.90
MDL number:
UNSPSC Code:
26111700
PubChem Substance ID:
NACRES:
NB.61

$523.00


Estimated to ship on15 April 2025


Request a Bulk Order

grade

for ion-selective electrodes

Quality Level

product line

Selectophore

mp

91-92 °C

SMILES string

CC(C)CN(CC(C)C)C(=S)SCc1ccccc1CSC(=S)N(CC(C)C)CC(C)C

InChI

1S/C26H44N2S4/c1-19(2)13-27(14-20(3)4)25(29)31-17-23-11-9-10-12-24(23)18-32-26(30)28(15-21(5)6)16-22(7)8/h9-12,19-22H,13-18H2,1-8H3

InChI key

JOEGUDLMKJATMX-UHFFFAOYSA-N

General description

Visit our Sensor Applications portal to learn more.

Application

Selected application examples

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Other Notes

Neutral ionophore for Cu2+-selective polymer membrane electrodes[1]

Legal Information

Selectophore is a trademark of Merck KGaA, Darmstadt, Germany

related product

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jinghui Li et al.
Analytica chimica acta, 1068, 11-17 (2019-05-11)
A novel freestanding all-solid-state polymeric membrane Cu2+-selective electrode (Cu2+-ISE) is proposed based on three-dimensional graphene sponge (3D GS). The 3D GS electrode has been characterized by cyclic voltammetry, electrochemical impedance spectroscopy, and contact angle measurements. Experiments show that 3D GS
S. Kamata et al.
Analyst, 114, 1029-1029 (1989)
Cemil Can Eylem et al.
Talanta, 183, 184-191 (2018-03-24)
A method was developed to determine copper and zinc simultaneously in brass sample using a multiple ion-selective electrode array system and multivariate calibration techniques. The multiple ion-selective electrode array system was consisted of one pH electrode and eight ion selective

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service