Skip to Content
Merck
All Photos(1)

Key Documents

60400

Sigma-Aldrich

Potassium iodide

puriss. p.a., ACS reagent, ≥99.0% (AT)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
KI
CAS Number:
Molecular Weight:
166.00
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
Assay:
≥99.0% (AT)
form:
solid
solubility:
water: soluble(lit.)

grade

ACS reagent
puriss. p.a.

vapor pressure

1 mmHg ( 745 °C)

Assay

≥99.0% (AT)

form

solid

impurities

≤0.001% total nitrogen (N)

loss

≤0.2% loss on drying, 150 °C

pH

6.0-9.2 (25 °C, 50 mg/mL in H2O)

mp

681 °C (lit.)

solubility

water: soluble(lit.)

anion traces

bromide, chloride (as Cl-): ≤100 mg/kg
iodate (IO3-): ≤3 mg/kg
phosphate (PO43-): ≤10 mg/kg
sulfate (SO42-): ≤50 mg/kg

cation traces

As: ≤0.1 mg/kg
Ba: ≤10 mg/kg
Ca: ≤10 mg/kg
Cd: ≤5 mg/kg
Co: ≤5 mg/kg
Cr: ≤5 mg/kg
Cu: ≤5 mg/kg
Fe: ≤2 mg/kg
Mg: ≤5 mg/kg
Mn: ≤5 mg/kg
Na: ≤50 mg/kg
Ni: ≤5 mg/kg
Pb: ≤5 mg/kg
Zn: ≤5 mg/kg

SMILES string

[K+].[I-]

InChI

1S/HI.K/h1H;/q;+1/p-1

InChI key

NLKNQRATVPKPDG-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Application

Potassium iodide is a metal halide, which is produced industrially, via treating potassium hydroxide with iodine or by reacting iron powder with iodine.[1] It is useful in treating the condition of dermatitis.[2]

Other Notes

A -D suffix exists for administrative purposes only.
All -D packages are 100% the same product, same quality, same specification as the package sizes previously sold without a -D.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

STOT RE 1 Oral

Target Organs

Thyroid

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Iodine Chemistry and Applications (2014)
Eagleson M.
Concise Encyclopedia Chemistry, 889-889 (1994)
Alumina-supported potassium iodide as a heterogeneous catalyst for biodiesel production from soybean oil.
Xie W and Li H.
J. Mol. Catal. A: Chem., 255(1), 1-9 (2006)
V Witko-Sarsat et al.
Journal of immunology (Baltimore, Md. : 1950), 161(5), 2524-2532 (1998-09-02)
We previously demonstrated the presence of advanced oxidation protein products (AOPP), a novel marker of oxidative stress in the plasma of uremic patients receiving maintenance dialysis. The present study in a cohort of 162 uremic patients showed that plasma concentrations
H B Li et al.
Journal of chromatography. A, 918(2), 335-339 (2001-06-16)
A novel method for the determination of iodide by size exclusion chromatography was established. The method was simple and highly sensitive with good precision. Iodide was converted to iodine, then sequestered with starch, and separated from the matrix using a

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service