Skip to Content
Merck
All Photos(1)

Documents

60032

Supelco

Potassium chloride

ready-to-use, traceable to BAM, in H₂O

Synonym(s):

Potassium chloride solution

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
KCl
CAS Number:
Molecular Weight:
74.55
Beilstein:
1711999
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:

product name

Potassium standard for AAS, ready-to-use, traceable to BAM, in H2O

shelf life

limited shelf life, expiry date on the label

concentration

in H2O
1.000 g/L K+

technique(s)

AAS: suitable

format

single component solution

SMILES string

[Cl-].[K+]

InChI

1S/ClH.K/h1H;/q;+1/p-1

InChI key

WCUXLLCKKVVCTQ-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Preparation Note

prepared with KCl and H2O

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shiqi Yang et al.
Scientific reports, 7(1), 13693-13693 (2017-10-24)
A five-year swine manure application trial and a study of nitrate leaching losses have been conducted. There were three treatments: traditional without manure (CK), traditional matched manure 4500 kg ha
Yonathan Gomez et al.
Biomedicines, 8(2) (2020-02-13)
A robust and easy-to-use tool for the ex vivo dynamic evaluation of pancreatic islet (PI) function is essential for further development of novel cell-based therapeutic approaches to treating diabetes. Here, we developed four different glucose perifusion protocols (GPPs) in a
Yu Yang et al.
Environmental science & technology, 48(11), 6391-6400 (2014-04-24)
Titanium dioxide (TiO2) is widely used in food products, which will eventually enter wastewater treatment plants and terrestrial or aquatic environments, yet little is known about the fraction of this TiO2 that is nanoscale, or the physical and chemical properties
Carlotta Pucci et al.
Soft matter, 10(48), 9657-9667 (2014-10-31)
The phase behavior of an ad hoc synthesized surfactant, sodium 8-hexadecylsulfate (8-SHS), and its mixtures with didecyldimethylammonium bromide (DiDAB) in water is reported. We dealt with dilute concentration regimes, at a total surfactant content of <30 mmol kg(-1) where vesicular
Jing Huang et al.
Applied and environmental microbiology, 80(1), 29-42 (2013-10-01)
Bacteria play important roles in mineral weathering and soil formation. However, few reports of mineral weathering bacteria inhabiting subsurfaces of soil profiles have been published, raising the question of whether the subsurface weathering bacteria are fundamentally distinct from those in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service