Skip to Content
Merck
All Photos(1)

Documents

60029

Supelco

Potassium standard for AAS

ready-to-use, in nitric acid

Synonym(s):

Potassium nitrate solution

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:

shelf life

limited shelf life, expiry date on the label

concentration

in nitric acid
1.000 g/L K+

technique(s)

AAS: suitable

format

single component solution

SMILES string

[K+].[O-][N+]([O-])=O

InChI

1S/K.NO3/c;2-1(3)4/q+1;-1

InChI key

FGIUAXJPYTZDNR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Preparation Note

prepared with KNO3 and HNO3 (~0.5 M)

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Met. Corr. 1 - Skin Irrit. 2

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Wolfgang R Engelsberger et al.
The Plant journal : for cell and molecular biology, 69(6), 978-995 (2011-11-09)
Nitrogen is an essential macronutrient for plant growth and development. Inorganic nitrogen and its assimilation products control various metabolic, physiological and developmental processes. Although the transcriptional responses induced by nitrogen have been extensively studied in the past, our work here
Mila Ivanova Luleva et al.
Sensors (Basel, Switzerland), 11(4), 4188-4206 (2011-12-14)
Displacement of soil particles caused by erosion influences soil condition and fertility. To date, the cesium 137 isotope ((137)Cs) technique is most commonly used for soil particle tracing. However when large areas are considered, the expensive soil sampling and analysis
Redrothu Hanumantharao et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 99, 181-188 (2012-10-16)
Nonlinear optical crystals of Bis (l-glutamine) potassium nitrate (BGPN) were grown by slow evaporation technique at ambient temperature. Solubility and metastable zone width of BGPN in aqueous solution were determined. The grown crystal was characterized by single crystal XRD, Powder
Changhong Yao et al.
Bioresource technology, 118, 438-444 (2012-06-22)
Microalgal starch is a potential feedstock for biofuel production. The effects of KNO(3) and MgSO(4) concentrations and light intensity on biomass and starch production by the marine microalga, Tetraselmis subcordiformis, were investigated. Under 200 μmol m(-2) s(-1) irradiance and sulfur-deprived
A R Pradeep et al.
Australian dental journal, 57(4), 429-434 (2012-11-29)
Dentinal hypersensitivity has been defined as a short, sharp pain arising from exposed dentine as a result of various stimuli such as heat, cold, chemical, or osmotic, that cannot be ascribed to any other pathology. This study was conducted to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service