Skip to Content
Merck
All Photos(1)

Key Documents

58170

Sigma-Aldrich

α-Ionone

technical, ≥90% (GC)

Synonym(s):

4-(2,6,6-Trimethyl-2-cyclohexenyl)-3-buten-2-one

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H20O
CAS Number:
Molecular Weight:
192.30
Beilstein:
3197885
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

grade

technical

Assay

≥90% (GC)

impurities

<10% β-ionone

refractive index

n20/D 1.498 (lit.)
n20/D 1.500

bp

259-263 °C (lit.)

density

0.93 g/mL at 25 °C (lit.)

SMILES string

CC(=O)\C=C\C1C(C)=CCCC1(C)C

InChI

1S/C13H20O/c1-10-6-5-9-13(3,4)12(10)8-7-11(2)14/h6-8,12H,5,9H2,1-4H3/b8-7+

InChI key

UZFLPKAIBPNNCA-BQYQJAHWSA-N

Looking for similar products? Visit Product Comparison Guide


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hao Cai et al.
Molecules (Basel, Switzerland), 18(2), 1368-1382 (2013-01-26)
Flos Lonicerae Japonicae (FLJ) is a popular herb used for many centuries in Traditional Chinese Medicine as a treatment of fever and inflammation. Non-fumigated processing of FLJ has been the traditional approach used in post-harvest preparation of the commodity for
Darunee Soorukram et al.
Organic letters, 6(14), 2409-2411 (2004-07-02)
[reaction: see text] The allylic substitution of sterically hindered (2-iodocycloalkyl)phosphates proceeds with complete anti S(N)2' stereoselectivity with mixed diorganozincs of the type RZnCH(2)SiMe(3) in the presence of CuCN.2LiCl. Only the group R of the copper-zinc reagent is transferred in the
Hye Gwang Jeong et al.
Archives of pharmacal research, 25(2), 197-201 (2002-05-15)
Beta-lonone has been reported to induce the cytochrome P450 (P450) 281 in rats. In this study, the effects of beta-ionone and an isomer, alpha-ionone, on liver P450 1A and 2B expression in Sprague Dawley rats were investigated. Subcutaneous administration of
Sabine Sewenig et al.
Journal of agricultural and food chemistry, 53(4), 838-844 (2005-02-17)
A new coupling system of GC-GC, connected via a Multi Column Switching Device MCS2 for measuring isotope ratios, is introduced. By means of several standard substances the precise and accurate measurement of isotopic values is proved. First applications concerning the
J Lalko et al.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association, 45 Suppl 1, S251-S257 (2007-11-23)
A toxicologic and dermatologic review of ionone when used as a fragrance ingredient is presented.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service