Skip to Content
Merck
All Photos(1)

Key Documents

56681

Supelco

Hydrocarbon Kit acc. to DIN EN 14039/ISO 16703

Synonym(s):

Alkane content in waste and soil according DIN EN 14039/ISO 16703, Kit, Mineral oil content in waste and soil according DIN EN 14039/ISO 16703, Kit, Waste hydrocarbon content in waste and soil according DIN EN 14039/ISO 16703, Kit

Sign Into View Organizational & Contract Pricing

Select a Size

1 KIT
$1,500.00

$1,500.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 KIT
$1,500.00

About This Item

UNSPSC Code:
41113319
NACRES:
NA.21

$1,500.00


Check Cart for Availability

Request a Bulk Order

grade

analytical standard

Agency

DIN

shelf life

limited shelf life, expiry date on the label

application(s)

environmental

Related Categories

Application

The hydrocarbon kit is intended to be used for the determination of hydrocarbons in solid waste, within a boiling point range of 175 °C to 525 °C, by gas chromatography and in accordance with DIN EN 14039 and ISO 16703. The hydrocarbons include n-alkanes from C10H22 to C40H82, isoalkanes, cycloalkanes, alkyl benzenes, alkyl napthalenes, and polycyclic aromatic compounds.

Components

kit contains:
mineral oil standard for calibration 8 mg/ml in heptane
alkane standard solution
heptane
standard solution for the determination of the retention time
Florisil® tubes

Legal Information

Florisil is a registered trademark of U.S. Silica Company

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Asp. Tox. 1 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

3 - Flammable liquids

Flash Point(F)

24.8 °F

Flash Point(C)

-4 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Protocols

Special mineral oil standard mixtures are gravimetrically produced for calibration.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service