Skip to Content
Merck
All Photos(1)

Key Documents

55736

Supelco

Orientin

analytical standard

Synonym(s):

8-β-D-Glucopyranosyl-3′,4′,5,7-tetrahydroxyflavone, Luteolin 8-C-β-D-glucopyranoside, Luteolin-8-glucoside, Lutexin

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H20O11
CAS Number:
Molecular Weight:
448.38
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:

grade

analytical standard

Quality Level

Assay

≥98.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

impurities

≤5.0% water

application(s)

food and beverages

format

neat

SMILES string

OC[C@H]1O[C@H]([C@H](O)[C@@H](O)[C@@H]1O)c2c(O)cc(O)c3C(=O)C=C(Oc23)c4ccc(O)c(O)c4

InChI

1S/C21H20O11/c22-6-14-17(28)18(29)19(30)21(32-14)16-11(26)4-10(25)15-12(27)5-13(31-20(15)16)7-1-2-8(23)9(24)3-7/h1-5,14,17-19,21-26,28-30H,6H2/t14-,17-,18+,19-,21+/m1/s1

InChI key

PLAPMLGJVGLZOV-VPRICQMDSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Orientin is a flavone existing in a variety of plants and exhibits a broad-spectrum of pharmacological properties such as free-radical inhibition, antiviral, radioprotection, anti-oxidant and vasorelaxant property.[1]

Application

Orientin may be used as an analytical reference standard for the quantification of the analyte in leaves of Cecropia species[2] and biological samples[1] using various chromatography techniques.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Biochem/physiol Actions

Flavonoid glycoside found in the acai tree and globe and passion flowers. Antiviral, antimicrobial and antioxidant. Exhibits radioprotective activities.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Determination and pharmacokinetics of orientin in rabbit plasma by liquid chromatography after intravenous administration of orientin and Trollius chinensis Bunge extract
Li X, et al.
Journal of Chromatography. B, Biomedical Sciences and Applications, 853(1-2), 221-226 (2007)
An HPLC-DAD method to quantification of main phenolic compounds from leaves of Cecropia species
Costa.MG, et al.
Journal of the Brazilian Chemical Society, 22(6), 1096-1102 (2011)
Shagufta Perveen et al.
Journal of Asian natural products research, 13(9), 799-804 (2011-08-13)
Two new C-glycosylflavonoids celtisides A (1) and B (2) have been isolated from n-butanol-soluble fraction of Celtis africana, along with five known C-glycosylflavonoids vitexin (3), orientin (4), isoswertiajaponin (5), isoswertisin (6), and 2″-O-rhamnosyl vitexin (7) reported for the first time
V Nayak et al.
Radiation research, 163(2), 165-171 (2005-01-22)
In a previous study, orientin and vicenin, the water-soluble plant flavonoids, protected mice against radiation lethality (Uma Devi et al., Radiat. Res. 151, 74-78, 1999). To study bone marrow protection, adult Swiss mice were exposed to 0-6 Gy 60Co gamma
Dalene de Beer et al.
Journal of the science of food and agriculture, 92(2), 274-282 (2011-07-23)
The effects of citric and ascorbic acids on the stability of aspalathin in rooibos (Aspalathus linearis) ready-to-drink (RTD) formulations containing fermented rooibos extract (FR), aspalathin-enriched green rooibos extract (GR) and aspalathin-enriched green rooibos extract ascorbic acid solubilisate (GR-solubilisate) were investigated

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service