Skip to Content
Merck
All Photos(1)

Key Documents

55695

Sigma-Aldrich

3-Hydroxy-3-methylglutaric acid

purum, ≥95.0% (GC)

Synonym(s):

3-Hydroxy-3-methylpentanedioic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOC(CH3)(CH2CO2H)2
CAS Number:
Molecular Weight:
162.14
Beilstein:
1769194
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

grade

purum

Assay

≥95.0% (GC)

mp

105-108 °C (lit.)
105-109 °C

SMILES string

CC(O)(CC(O)=O)CC(O)=O

InChI

1S/C6H10O5/c1-6(11,2-4(7)8)3-5(9)10/h11H,2-3H2,1H3,(H,7,8)(H,9,10)

InChI key

NPOAOTPXWNWTSH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

replaced by

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mohamad Eid Hammadeh et al.
American journal of reproductive immunology (New York, N.Y. : 1989), 47(2), 82-90 (2002-03-20)
The aims of the present study were to (i) determine the presence and concentration of albumin fractions (alpha1, alpha2, beta, gamma), immunoglobulins (IgA, IgG, IgM) and cytokines [interleukin (IL)-6, IL-8, granulocyte-macrophage colony-stimulating factor (GM-CSF)] in periovulatory ovarian follicular fluid (FF)
Alister D Muir
Journal of AOAC International, 89(4), 1147-1157 (2006-08-19)
Flaxseed (Linum usitatissimum L.) is a major source of dietary intake of lignans by virtue of the high concentrations (0.7-1.5%) that are present in the seed. The principal lignan present in flaxseed is secoisolariciresinol diglucoside (SDG), which occurs as a
S Funghini et al.
Molecular genetics and metabolism, 73(3), 268-275 (2001-07-20)
3-Hydroxy-3-methylglutaric aciduria is a rare autosomal recessive inborn error of metabolism caused by deficiency of the mitochondrial enzyme 3-hydroxy-3-methylglutaryl-CoA lyase (HMGCL). Up to now only a few mutations have been reported in the HMGCL gene. We report the first Italian
C Mir et al.
Journal of inherited metabolic disease, 29(1), 64-70 (2006-04-08)
3-Hydroxy-3-methylglutaric aciduria is a rare autosomal recessive genetic disorder that affects ketogenesis and leucine metabolism. The disease is caused by mutations in the gene coding for 3-hydroxy-3-methylglutaryl-coenzyme A lyase (HL). To date 26 different mutations have been described. A (betaalpha)(8)
Elaine A Porter et al.
Phytochemistry, 81, 90-96 (2012-06-23)
LC-UV-MS/MS analysis of leaf extracts from 146 accessions of 71 species of Rosa revealed that some taxa accumulated flavonol O-glycosides acylated with 3-hydroxy-3-methylglutaric acid, which are relatively uncommon in plants. The structures of two previously unrecorded examples isolated from Rosa

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service