Skip to Content
Merck
All Photos(1)

Key Documents

55639

Supelco

Tebuconazole-(tert-butyl-d9)

PESTANAL®, analytical standard

Synonym(s):

Tebuconazole-d9

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$645.00

$645.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
$645.00

About This Item

Empirical Formula (Hill Notation):
C16D9H13ClN3O
CAS Number:
Molecular Weight:
316.87
MDL number:
UNSPSC Code:
77101502
PubChem Substance ID:
NACRES:
NA.24

$645.00


Check Cart for Availability

Request a Bulk Order

grade

analytical standard

Quality Level

isotopic purity

≥97 atom % D

product line

PESTANAL®

Assay

≥98.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

application(s)

agriculture
environmental

format

neat

SMILES string

ClC(C=C1)=CC=C1CCC(C(C([2H])([2H])[2H])(C([2H])([2H])[2H])C([2H])([2H])[2H])(O)CN2N=CN=C2

InChI

1S/C16H22ClN3O/c1-15(2,3)16(21,10-20-12-18-11-19-20)9-8-13-4-6-14(17)7-5-13/h4-7,11-12,21H,8-10H2,1-3H3/i1D3,2D3,3D3

InChI key

PXMNMQRDXWABCY-GQALSZNTSA-N

Related Categories

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 2 - Repr. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 3

1 of 3

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service