Skip to Content
Merck
All Photos(1)

Key Documents

55524

Supelco

1-Decen-3-ol

analytical standard

Synonym(s):

3-Hydroxy-1-decene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)6CH(OH)CH=CH2
CAS Number:
Molecular Weight:
156.27
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:

grade

analytical standard

Assay

≥98.0% (GC)

shelf life

limited shelf life, expiry date on the label

refractive index

n20/D 1.444 (lit.)
n20/D 1.444

bp

214-217 °C (lit.)

density

0.837 g/mL at 25 °C (lit.)

application(s)

cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care

SMILES string

CCCCCCCC(O)C=C

InChI

1S/C10H20O/c1-3-5-6-7-8-9-10(11)4-2/h4,10-11H,2-3,5-9H2,1H3

InChI key

NSFWLHKFTGZFBP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

197.0 °F - closed cup

Flash Point(C)

91.67 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service