Skip to Content
Merck
All Photos(2)

Documents

54746

Supelco

Mefluidide

PESTANAL®, analytical standard

Synonym(s):

5′-(1,1,1-Trifluoromethanesulfonamido)acet-2′,4′-xylidide, 5-Acetamido-2,4-dimethyltrifluoromethanesulfonanilide, N-[2,4-Dimethyl-5-[[(trifluoromethyl)sulfonyl]amino]phenyl]acetamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H13F3N2O3S
CAS Number:
Molecular Weight:
310.29
Beilstein:
2819120
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

Assay

≥98.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

composition

carbon content, 41.73-43.43%

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

SMILES string

CC(=O)Nc1cc(NS(=O)(=O)C(F)(F)F)c(C)cc1C

InChI

1S/C11H13F3N2O3S/c1-6-4-7(2)10(5-9(6)15-8(3)17)16-20(18,19)11(12,13)14/h4-5,16H,1-3H3,(H,15,17)

InChI key

OKIBNKKYNPBDRS-UHFFFAOYSA-N

General description

Mefluidide is a herbicide widely used to regulate the growth of turf grass and ornamentals. Its primary use is to inhibit cell division in meristematic regions of the plant.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Development of a multiresidue method for nitrogen-containing pesticides
Baumann.A.R, et al.
Fresenius Journal of Analytical Chemistry, 339(6), 357-364 (1991)
S K Wimer et al.
Journal of animal science, 63(4), 1054-1062 (1986-10-01)
Mefluidide, a plant growth regulator, was evaluated in two cow-calf grazing trials and one herbage trial on smooth brome (Bromus inermis) pastures stocked at recommended densities in eastern Nebraska. Mefluidide-treated pasture increased cow and calf production during August of 1982
K E Turner et al.
Journal of animal science, 68(10), 3399-3405 (1990-10-01)
Twelve Hereford heifers were used in a grazing experiment to determine intake and digestibility of tall fescue forage treated with the plant growth regulator mefluidide. Additionally, steer and heifer performance were evaluated after grazing tall fescue pastures or consuming hay
Fate of the plant growth regulator mefluidide [N-[2,4-dimethyl-5-[[(trifluoromethyl)sulfonyl]amino]phenyl]acetamide] in a cow and sheep.
G W Ivie
Journal of agricultural and food chemistry, 28(6), 1286-1288 (1980-11-01)
Poa Annua: Physiology, Culture, and Control of Annual Bluegrass, 339(6), 357-364 (2003)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service