Skip to Content
Merck
All Photos(1)

Documents

54730

Supelco

6-Acetyl-1,1,2,4,4,7-hexamethyltetralin

analytical standard

Synonym(s):

1,2,3,4-Tetrahydro-1,1,3,4,4,6-hexamethyl-7-naphthyl methyl ketone, 1-(5,6,7,8-Tetrahydro-3,5,5,6,8,8-hexamethyl-2-naphthalenyl)ethanone, 5′,6′,7′,8′-Tetrahydro-3′,5′,5′,6′,8′,8′-hexamethyl-2′-acetonaphthone, 6-Acetyl-1,1,2,4,4,7-hexamethyl-1,2,3,4-tetrahydronaphthalene, AHMT, Fixolide, Musk tonalid, NSC 19550

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H26O
CAS Number:
Molecular Weight:
258.40
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥97.0% (GC)

shelf life

limited shelf life, expiry date on the label

impurities

≤1.0% water

application(s)

cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care

format

neat

SMILES string

CC1CC(C)(C)c2cc(C(C)=O)c(C)cc2C1(C)C

InChI

1S/C18H26O/c1-11-8-16-15(9-14(11)13(3)19)17(4,5)10-12(2)18(16,6)7/h8-9,12H,10H2,1-7H3

InChI key

DNRJTBAOUJJKDY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

6-Acetyl-1,1,2,4,4,7-hexamethyltetralin (musk tonalide) is a polycyclic musk which belongs to the class of tetralin compounds. It is widely used as a fragrance ingredient in consumer products, mainly soaps, detergents, cosmetics, and cleaning agents.

Application

6-Acetyl-1,1,2,4,4,7-hexamethyltetralin may be used as an analytical reference standard for the determination of the analyte in water samples, fish and perfumes by chromatography-based techniques.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Gas chromatography-mass spectrometry screening methods for select UV filters, synthetic musks, alkylphenols, an antimicrobial agent, and an insect repellent in fish
Mottaleb MA, et al.
Journal of Chromatography A, 1216(5), 815-823 (2009)
Examination of the in vitro (anti) estrogenic,(anti) androgenic and (anti) dioxin-like activities of tetralin, indane and isochroman derivatives using receptor-specific bioassays
Schreurs Richard HMM, et al.
Toxicology Letters, 156(2), 261-275 (2005)
Chromogenic derivatives of AHTN (6-acetyl-1, 1, 2, 4, 4, 7-hexamethyltetralin) react with amino acids and protein in vitro. Spectral characteristics of the colour products
Pipino S, et al.
Food And Chemical Toxicology, 42(5), 785-790 (2004)
Multicomponent analytical methodology to control phthalates, synthetic musks, fragrance allergens and preservatives in perfumes
Sanchez-Prado L, et al.
Talanta, 85(1), 370-379 (2011)
Semi-automated hollow-fibre membrane extraction, a novel enrichment technique for the determination of biologically active compounds in water samples
Muller S, et al.
Journal of Chromatography A, 985(1-2), 99-106 (2003)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service