Skip to Content
Merck
All Photos(1)

Key Documents

53777

Millipore

HybriScan Wash Solution

suitable for microbiology, For use with HybriScan

Synonym(s):

Wash buffer

Sign Into View Organizational & Contract Pricing

Select a Size

500 ML
$108.00

$108.00


Estimated to ship on28 April 2025


Request more information

Select a Size

Change View
500 ML
$108.00

About This Item

UNSPSC Code:
41171621
NACRES:
NA.85

$108.00


Estimated to ship on28 April 2025


Request more information

Quality Level

feature

ready-to-use

manufacturer/tradename

(HybriScan)

technique(s)

microbe id | in situ hybriziation: suitable

capacity

90 mL

volume

90 mL

application(s)

microbiology

storage temp.

2-8°C

General description

HybriScan Washing Solution is designed to be used with the HybriScan detection system. The HybriScan system is based on the detection of microbe-specific rRNA using sandwich hybridization. The signal read-out is triggered optically by an enzymatically-generated color change. The results are available within 2 hours of testing with a pre-enrichment step. Sandwich hybridization provides sensitivity in crude biological samples because it is not susceptible to matrix interference. By using specific probes, the HybriScan system allows both genus and species-specific detection. The wash solution is used to remove any unbound molecules like probes and enzymes before the enzyme-substrate reaction.

Application

HybriScan Washing Solution is used with the HybriScan microbial detection system based on sandwich hybridization in many analytical fields, including monitoring the microbial content of beer, wine, non-alcoholic beverages, drinking water, a wide variety of foods, and wastewater.

Legal Information

HybriScan is a trademark of ScanBec GmbH

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service