Skip to Content
Merck
All Photos(2)

Key Documents

51872

Supelco

Piroctone olamine

analytical standard, PESTANAL®

Synonym(s):

1-Hydroxy-4-methyl-6-(2,4,4-trimethylpentyl)-2(1H)-pyridone ethanolammonium salt

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
$203.00

$203.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 MG
$203.00

About This Item

Empirical Formula (Hill Notation):
C14H23NO2 · C2H7NO
CAS Number:
Molecular Weight:
298.42
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

$203.00


Check Cart for Availability

Request a Bulk Order

grade

analytical standard

Quality Level

product line

PESTANAL®

Assay

≥99.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

storage temp.

2-8°C

SMILES string

NCCO.CC(CC1=CC(C)=CC(=O)N1O)CC(C)(C)C

InChI

1S/C14H23NO2.C2H7NO/c1-10-6-12(15(17)13(16)8-10)7-11(2)9-14(3,4)5;3-1-2-4/h6,8,11,17H,7,9H2,1-5H3;4H,1-3H2

InChI key

BTSZTGGZJQFALU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Piroctone olamine is a pyridone derivative, which is known to have bactericidal effects on gram-positive and gram-negative bacteria and fungicidal effects and hence is a component of many cosmetic products such as anti-dandruff shampoo.[1]

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Eye Dam. 1 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

323.4 °F - Pensky-Martens closed cup

Flash Point(C)

161.9 °C - Pensky-Martens closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tiziana Di Marco et al.
Cancer letters, 476, 106-119 (2020-02-18)
The coatomer protein complex zeta 1 (COPZ1) represents a non-oncogene addiction for thyroid cancer (TC); its depletion impairs the viability of thyroid tumor cells, leads to abortive autophagy, ER stress, UPR and apoptosis, and reduces tumor growth of TC xenograft
Increased in vivo efficacy of lenalidomide by addition of piroctone olamine
Kim Y
In Vivo, 25, 99-103 (2011)
Ilona Wall et al.
Anticancer research, 34(10), 5375-5380 (2014-10-03)
Activated Wnt signaling in cancer cells leads to cell proliferation. It has been shown that the Wnt pathway is activated in pancreatic adenocarcinoma cells. Therefore, we tested the effect of Wnt inhibitors in human and murine pancreatic cancer cell lines.
Viktoryia Sidarovich et al.
Molecular pharmacology, 87(3), 513-524 (2015-01-08)
Iron is an essential cellular nutrient, being a critical cofactor of several proteins involved in cell growth and replication. Compared with normal cells, neoplastic cells have been shown to require a greater amount of iron, thus laying the basis for

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service