Skip to Content
Merck
All Photos(1)

Key Documents

51706

Supelco

QC-Standard Solution for EN ISO 9377-2

analytical standard

Sign Into View Organizational & Contract Pricing

Select a Size

1 ML
$146.00

$146.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 ML
$146.00

About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

$146.00


Check Cart for Availability

Request a Bulk Order

grade

analytical standard

Quality Level

Agency

EN 9377-2
ISO

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

environmental
petroleum

format

multi-component solution

Related Categories

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Components

contains Mineral oil type A and B ~5 mg/ml each, BAM reference material 5002 and 5003 in n-Heptane

Signal Word

Danger

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Asp. Tox. 1 - Carc. 2 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

-0.4 °F - closed cup

Flash Point(C)

-18 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 2

1 of 2

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service