Skip to Content
Merck
All Photos(1)

Documents

51254

Supelco

Protein Quantification Kit-Rapid

sufficient for 500 tests

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12164500
NACRES:
NA.21

usage

sufficient for 500 tests

Quality Level

analyte chemical class(es)

proteins

technique(s)

photometry: suitable

storage temp.

2-8°C

Application

This kit enables precise protein determination within a few minutes.

Components

Kit contains: BSA Standard solution (4.00 mg/ml) 1.5 mL × 1 vial, Coomassie, Brilliant Blue G solution 100 mL × 2 bottles, protocol

Legal Information

Pictograms

Corrosion

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Met. Corr. 1 - Skin Irrit. 2

Storage Class Code

8A - Combustible corrosive hazardous materials

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chenteng Lai et al.
Molecular and cellular biochemistry (2022-08-25)
MicroRNAs (miRs) are regulators of number of cellular process. miRs enclosed within exosomes can be crucial regulators of intercellular signalling and could be an important biomarker of various age-associated disorders. Role of exosomal enclosed miRs in osteoarthritis (OA) chondrocytes and
Nan Wang et al.
International journal of molecular medicine, 39(4), 783-790 (2017-02-17)
Fisetin (3,3',4',7-tetrahydroxyflavone) is a plant flavonol found in fruits and vegetables that has been reported to inhibit migration and proliferation in several types of cancer. Reactive astrogliosis involves astrocyte migration and proliferation, and contributes to the formation of glial scars
Protective effects of radon inhalation on carrageenan-induced inflammatory paw edema in mice.
Inflammation, 35.2, 713-722 (2012)
Diana Vilela et al.
ACS applied materials & interfaces, 13(13), 14964-14973 (2021-03-27)
The low efficacy of current conventional treatments for bacterial infections increases mortality rates worldwide. To alleviate this global health problem, we propose drug-free enzyme-based nanomotors for the treatment of bacterial urinary-tract infections. We develop nanomotors consisting of mesoporous silica nanoparticles
M M M Kandiel et al.
Iranian journal of veterinary research, 19(4), 270-275 (2019-02-19)
Egypt is anticipated to be potentially influenced by the global climate warming. Therefore, the current study aimed to evaluate the influence of age on the fertility potential of Arabian stallions during summer breeding months. Arabian horses grouped according to their

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service