Skip to Content
Merck
All Photos(1)

Key Documents

50797

Supelco

Hexylene glycol

analytical standard

Synonym(s):

(±)-2-Methyl-2,4-pentanediol, MPD

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH(OH)CH2C(CH3)2OH
CAS Number:
Molecular Weight:
118.17
Beilstein:
1098298
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

vapor density

4.1 (vs air)

vapor pressure

0.02 mmHg ( 20 °C)

Assay

≥99.0% (GC)

shelf life

limited shelf life, expiry date on the label

expl. lim.

7.4 %

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

refractive index

n20/D 1.427 (lit.)
n20/D 1.427-1.429

bp

197 °C (lit.)

mp

−40 °C (lit.)

density

0.925 g/mL at 25 °C (lit.)

application(s)

cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care

format

neat

SMILES string

CC(O)CC(C)(C)O

InChI

1S/C6H14O2/c1-5(7)4-6(2,3)8/h5,7-8H,4H2,1-3H3

InChI key

SVTBMSDMJJWYQN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

201.2 °F - closed cup

Flash Point(C)

94 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

D Spoerl et al.
Dermatology (Basel, Switzerland), 220(3), 238-242 (2010-02-27)
We report a case of severe contact urticaria with systemic involvement resembling an anaphylactic reaction, following the application of a topical corticosteroid. This was caused by hexylene glycol, an excipient in the formulation. Glycols are widely used in cosmetics, foods
Huan Shu et al.
Genome biology, 15(4), R62-R62 (2014-04-09)
Histone variants establish structural and functional diversity of chromatin by affecting nucleosome stability and histone-protein interactions. H3.3 is an H3 histone variant that is incorporated into chromatin outside of S-phase in various eukaryotes. In animals, H3.3 is associated with active
J Xu et al.
Cell death & disease, 6, e1616-e1616 (2015-01-23)
The selenoprotein thioredoxin reductase 1 (TrxR1) has several key roles in cellular redox systems and reductive pathways. Here we discovered that an evolutionarily conserved and surface-exposed tryptophan residue of the enzyme (Trp114) is excessively reactive to oxidation and exerts regulatory
Yu Zhu et al.
Molecular pharmaceutics, 12(2), 332-341 (2014-08-26)
Polycations are explored as carriers to deliver therapeutic nucleic acids. Polycations are conventionally pharmacological inert with the sole function of delivering therapeutic cargo. This study reports synthesis of a self-immolative polycation (DSS-BEN) based on a polyamine analogue drug N(1),N(11)-bisethylnorspermine (BENSpm).
Umile Gianfranco Spizzirri et al.
Pharmaceutical development and technology, 20(3), 288-296 (2013-12-03)
In this study, pH-responsive hydrogels, synthesized by the coupling reaction of polyacrylic acid and catechin, are proposed as carriers of oxidable drugs toward the GI tract. The presence of polyphenolic moieties in the network gives the polymers properties suitable for

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service