Skip to Content
Merck
All Photos(1)

Key Documents

49601

Sigma-Aldrich

L-Glutamic acid monopotassium salt monohydrate

puriss. p.a., ≥99.0%

Synonym(s):

Potassium L-glutamate

Sign Into View Organizational & Contract Pricing

Select a Size

100 G
$99.80
500 G
$296.00

$99.80


Available to ship on07 April 2025Details


Request a Bulk Order

Select a Size

Change View
100 G
$99.80
500 G
$296.00

About This Item

Linear Formula:
KOOCCH2CH2CH(NH2)COOH · H2O
CAS Number:
Molecular Weight:
203.23
Beilstein:
3637377
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.32

$99.80


Available to ship on07 April 2025Details


Request a Bulk Order

grade

puriss. p.a.

Assay

≥99.0% (NT)
≥99.0%

form

solid

optical activity

[α]20/D +23±1°, c = 5% in 5 M HCl

impurities

≤0.5% foreign amino acids

anion traces

chloride (Cl-): ≤100 mg/kg
sulfate (SO42-): ≤100 mg/kg

cation traces

Cd: ≤5 mg/kg
Co: ≤5 mg/kg
Cr: ≤5 mg/kg
Cu: ≤5 mg/kg
Fe: ≤5 mg/kg
Mg: ≤5 mg/kg
Mn: ≤5 mg/kg
NH4+: ≤200 mg/kg
Na: ≤500 mg/kg
Ni: ≤5 mg/kg
Pb: ≤5 mg/kg
Zn: ≤5 mg/kg

SMILES string

[K+].[H]O[H].N[C@@H](CCC([O-])=O)C(O)=O

InChI

1S/C5H9NO4.K.H2O/c6-3(5(9)10)1-2-4(7)8;;/h3H,1-2,6H2,(H,7,8)(H,9,10);;1H2/q;+1;/p-1/t3-;;/m0../s1

InChI key

XIBUKSQTWSKJMQ-QTNFYWBSSA-M

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lukas Aufinger et al.
Nature communications, 13(1), 2852-2852 (2022-05-24)
Complex dynamics such as period doubling and chaos occur in a wide variety of non-linear dynamical systems. In the context of biological circadian clocks, such phenomena have been previously found in computational models, but their experimental study in biological systems
Abdullah Ozer et al.
Nature protocols, 10(8), 1212-1233 (2015-07-17)
Because RNA-protein interactions have a central role in a wide array of biological processes, methods that enable a quantitative assessment of these interactions in a high-throughput manner are in great demand. Recently, we developed the high-throughput sequencing-RNA affinity profiling (HiTS-RAP)
Lipopolysaccharide increases bitter taste sensitivity via epigenetic changes in Tas2r gene clusters.
Lin, et al.
iScience, 26, 106920-106920 (2023)
Jin Zhang et al.
Cell, 179(2), 392-402 (2019-09-24)
The ability to sense sour provides an important sensory signal to prevent the ingestion of unripe, spoiled, or fermented foods. Taste and somatosensory receptors in the oral cavity trigger aversive behaviors in response to acid stimuli. Here, we show that

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service