Skip to Content
Merck
All Photos(1)

Key Documents

46345

Sigma-Aldrich

Fiske-Subbarow reducer

Synonym(s):

Fiske-Subbarow reagent

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352302
NACRES:
NA.25
Pricing and availability is not currently available.

Quality Level

Looking for similar products? Visit Product Comparison Guide

General description

formulation of 1-amino-2-naphthol-4-sulfonic acid, sodium sulfite and bisulfite according to C.H. Fiske and Y. Subbarow, J.Biol.Chem. 66, 375 (1925)

Application

The Fiske-Subbarow assay is a colorimetric assay designed to measure levels of inorganic phosphate in biological samples. Fiske-Subbarow reducer reagent is used as the reducer component of the Fiske-Subbarow assay.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1

Supplementary Hazards

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

E Durward et al.
BioTechniques, 25(4), 608-612 (1998-10-30)
Methods for detecting and measuring the success of nucleic acid sequence amplifications can be developed by detecting the by-products of amplification procedures. One method includes the detection of inorganic phosphate (Pi) during or on completion of the PCR. The method
A Kyaw et al.
Analytical biochemistry, 145(2), 230-234 (1985-03-01)
A new colorimetric procedure is described for inorganic phosphate determination using the color reaction between inorganic phosphate and acidified ammonium molybdate in the presence of Triton X-100. The method is simple and specific, and produces results comparable with those of
Eduardo H S Sousa et al.
Analytical biochemistry, 348(1), 57-63 (2005-11-18)
We describe the development of a rapid colorimetric assay for soluble guanylate cyclase (sGC) activity adapted for a 96-well microplate. The assay greatly decreases the analysis time and cost over traditional methodologies based on radio- and immunoassays and high-performance liquid
The colorimetric determination of phosphorus
Fiske, C. H., Subbarow, Y.
The Journal of Biological Chemistry, 66, 375-375 (1925)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service