Skip to Content
Merck
All Photos(1)

Documents

46341

Supelco

Imidacloprid solution

100 μg/mL in acetonitrile, PESTANAL®, analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H10ClN5O2
CAS Number:
Molecular Weight:
255.66
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

concentration

100 μg/mL in acetonitrile

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

single component solution

storage temp.

2-8°C

SMILES string

[O-][N+](=O)NC1=NCCN1Cc2ccc(Cl)nc2

InChI

1S/C9H10ClN5O2/c10-8-2-1-7(5-12-8)6-14-4-3-11-9(14)13-15(16)17/h1-2,5H,3-4,6H2,(H,11,13)

InChI key

YWTYJOPNNQFBPC-UHFFFAOYSA-N

General description

Imidacloprid is a neonicotinoid insecticide, which belongs to the chloronicotinyl nitroguanidine family. It is widely used to control crop pests and pet fleas.

Find more information here: Neonicotinoids

Application

Imidacloprid may be used as a reference standard for the determination of imidacloprid in cereal and vegetables by high-performance liquid chromatography method.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

related product

Product No.
Description
Pricing

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 2 - Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

35.6 °F - closed cup

Flash Point(C)

2 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Aston L Arthur et al.
Journal of economic entomology, 113(4), 1940-1945 (2020-04-30)
Lucerne flea (Sminthurus viridis Linnaeus) is an important establishment pest of winter grain crops and pastures in Australia. Control of S. viridis largely relies on the application of insecticides through foliar sprays or seed treatments; however, in recent years, farmers
HPLC determination of the new insecticide imidacloprid and its behavior in rice and cucumber.
Ishii Y, et al.
Journal of Agricultural and Food Chemistry, 42(12), 2917-2921 (1994)
Thomas G Bean et al.
Environmental science & technology, 53(7), 3888-3897 (2019-02-26)
Birds are potentially exposed to neonicotinoid insecticides by ingestion of coated seeds during crop planting. Adult male Japanese quail were orally dosed with wheat seeds coated with an imidacloprid (IMI) formulation at either 0.9 or 2.7 mg/kg body weight (BW)
Paul A Umina et al.
Experimental & applied acarology, 79(3-4), 345-357 (2019-11-11)
The redlegged earth mite (Halotydeus destructor) is an important agricultural pest in Australia, with a wide range of plant hosts. Halotydeus destructor has developed resistance to pyrethroid and organophosphorus insecticides as a consequence of the widespread use of these chemicals
Accidental human poisoning with a neonicotinoid insecticide, imidacloprid: A rare case report from rural India with a brief review of literature.
Kumar A, et al.
Egyptian Journal of Forensic Sciences, 3(4), 123-126 (2013)

Protocols

Learn more about Neonicotinoids - active substances used in plant protection products to control harmful insects.

Analysis of banned neonicotinoid insecticides from dandelion blossoms using QuEChERS and LC-MS.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service