Skip to Content
Merck
All Photos(1)

Key Documents

46309

Supelco

Griseofulvin

VETRANAL®, analytical standard

Synonym(s):

(2S)-trans-7-Chloro-2′,4,6-trimethoxy-6′-methylspiro(benzofuran-2[3H],1′-[2]cyclohexene)-3,4′-dione

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H17ClO6
CAS Number:
Molecular Weight:
352.77
Beilstein:
95226
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:

grade

analytical standard

product line

VETRANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

antibiotic activity spectrum

fungi

application(s)

agriculture
clinical
environmental
forensics and toxicology
pharmaceutical (small molecule)

format

neat

storage temp.

−20°C

SMILES string

ClC1=C(O[C@@]2(C(OC)=CC(C[C@H]2C)=O)C3=O)C3=C(OC)C=C1OC

InChI

1S/C17H17ClO6/c1-8-5-9(19)6-12(23-4)17(8)16(20)13-10(21-2)7-11(22-3)14(18)15(13)24-17/h6-8H,5H2,1-4H3/t8-,17+/m1/s1

InChI key

DDUHZTYCFQRHIY-RBHXEPJQSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

VETRANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Carc. 2 - Repr. 1B - Skin Sens. 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nidhi Aggarwal et al.
Colloids and surfaces. B, Biointerfaces, 105, 158-166 (2013-01-30)
The main objective of the study was to develop a microemulsion (ME) formulation of griseofulvin for the treatment of dermatophytosis (Indian Patent Application 208/DEL/2009). The oil phase was selected on the basis of drug solubility whereas the surfactant and cosurfactant
Marc S Raab et al.
Cancer research, 72(20), 5374-5385 (2012-09-04)
In contrast to normal cells, malignant cells are frequently aneuploid and contain multiple centrosomes. To allow for bipolar mitotic division, supernumerary centrosomes are clustered into two functional spindle poles in many cancer cells. Recently, we have shown that griseofulvin forces
Aditya K Gupta et al.
Pediatric dermatology, 30(1), 1-6 (2012-09-22)
Two oral antifungal agents, griseofulvin and terbinafine, have regulatory approval in the United States, but it is unknown whether one has superior overall efficacy. Genus-specific differences in efficacy are believed to exist for the two agents. It is not clear
S Knasmüller et al.
Critical reviews in toxicology, 27(5), 495-537 (1997-11-05)
Griseofulvin (GF) has been in use for more than 30 years as a pharmaceutical drug in humans for the treatment of dermatomycoses. Animal studies give clear evidence that it causes a variety of acute and chronic toxic effects, including liver
Mads H Rønnest et al.
Journal of medicinal chemistry, 52(10), 3342-3347 (2009-05-01)
Griseofulvin was identified as an inhibitor of centrosomal clustering in a recently developed assay. Centrosomal clustering is an important cellular event that enables bipolar mitosis for cancer cell lines harboring supernumerary centrosomes. We report herein the synthesis and SAR of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service