Skip to Content
Merck
All Photos(1)

Key Documents

46301

Supelco

Fluquinconazole

PESTANAL®, analytical standard

Synonym(s):

3-(2,4-Dichlorophenyl)-6-fluoro-2-(1H-1,2,4-trizol-1-yl)quinazolin-4(3H)-one, Castellan

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
$296.00

$296.00


Estimated to ship on02 June 2025


Request a Bulk Order

Select a Size

Change View
100 MG
$296.00

About This Item

Empirical Formula (Hill Notation):
C16H8Cl2FN5O
CAS Number:
Molecular Weight:
376.17
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

$296.00


Estimated to ship on02 June 2025


Request a Bulk Order

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

SMILES string

Fc1ccc2N=C(N(C(=O)c2c1)c3ccc(Cl)cc3Cl)n4cncn4

InChI

1S/C16H8Cl2FN5O/c17-9-1-4-14(12(18)5-9)24-15(25)11-6-10(19)2-3-13(11)22-16(24)23-8-20-7-21-23/h1-8H

InChI key

IJJVMEJXYNJXOJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Fluquinconazole is a triazole, which is used as a fungicide for foliar applications.[1] It is widely used in cereal seed treatment since it controls the root-rot pathogen such as Gaeumannomyces graminis, protects against infection by Puccinia spp. and Septoria spp. leaf diseases and helps in controlling seedborne diseases such as Tilletia spp. and Ustilago spp.

Application

Fluquinconazole may be used as a reference standard in the determination of fluquinconazole in liquid and fruit samples using solid-phase microextraction followed by high-performance liquid chromatography coupled with diode array detector (SPME-HPLC-DAD).[2]
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Acute Tox. 4 Dermal - Aquatic Acute 1 - Aquatic Chronic 1 - Skin Irrit. 2 - STOT RE 1 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sensitivity of Venturia inaequalis to five DMI fungicides, including the new triazole fluquinconazole, and to pyrimethanil.
Schnabel G and Parisi L
Journal of Plant Diseases and Protection, 36-46 (1997)
Seed treatment with fluquinconazole for control of cereal take-all, foliar and seed-borne diseases.
Brighton Crop Protection Conference: Pests & Diseases, 907-912 (1998)
Sensitivity of Venturia inaequalis to five DMI fungicides, including the new triazole fluquinconazole, and to pyrimethanil.
Schnabel G and Parisi L
Journal of Plant Diseases and Protection, 36-46 (1997)
Development and application of a screening method for triazole fungicide determination in liquid and fruit samples using solid-phase microextraction and HPLC-DAD.
Bordagaray A, et al.
Analytical Methods : Advancing Methods and Applications, 5(10), 2565-2571 (2013)
Angela P Van de Wouw et al.
PloS one, 12(11), e0188106-e0188106 (2017-11-16)
Leptosphaeria maculans is the major pathogen of canola (oilseed rape, Brassica napus) worldwide. In Australia, the use of azole fungicides has contributed to the 50-fold increase in canola production in the last 25 years. However, extensive application of fungicides sets

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service