Skip to Content
Merck
All Photos(1)

Key Documents

46204

Supelco

4-Heptanone

analytical standard

Synonym(s):

Butyrone, Dipropyl ketone

Sign Into View Organizational & Contract Pricing

Select a Size

5 ML
$257.00

$257.00


Estimated to ship on02 June 2025


Request a Bulk Order

Select a Size

Change View
5 ML
$257.00

About This Item

Linear Formula:
(CH3CH2CH2)2CO
CAS Number:
Molecular Weight:
114.19
Beilstein:
1699049
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

$257.00


Estimated to ship on02 June 2025


Request a Bulk Order

grade

analytical standard

vapor density

3.93 (vs air)

vapor pressure

5.2 mmHg ( 20 °C)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

refractive index

n20/D 1.407 (lit.)

bp

145 °C (lit.)

mp

−33 °C (lit.)

density

0.817 g/mL at 25 °C (lit.)

application(s)

cleaning products
cosmetics
environmental
flavors and fragrances
food and beverages
personal care

format

neat

SMILES string

CCCC(=O)CCC

InChI

1S/C7H14O/c1-3-5-7(8)6-4-2/h3-6H2,1-2H3

InChI key

HCFAJYNVAYBARA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

4-Heptanone may be used as an analytical reference standard for the quantification of the analyte in cane-sugar spirit and rum samples using high-performance liquid chromatography with diode array detection.[1]
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

120.2 °F - closed cup

Flash Point(C)

49 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

T H Gan et al.
Analytical chemistry, 72(4), 807-815 (2000-03-04)
The ion mobility (IMS) spectra of the alkanolamines, monoethanolamine (MEA), 3-amino-1-propanol (PRA), 4-amino-1-butanol (BUA), and 5-amino-1-pentanol (PEA) with acetone and 4-heptanone reagent gases have been measured using a hand-held spectrometer. Monomer and dimer peak patterns were observed for all the
Hans Günther Wahl et al.
Nephrology, dialysis, transplantation : official publication of the European Dialysis and Transplant Association - European Renal Association, 19(10), 2576-2583 (2004-07-29)
There is an ongoing discussion about the risks of di(2-ethylhexyl) phthalate (DEHP) exposure for the general population as well as for specific subgroups in various medical settings. Haemodialysis patients certainly belong to the group with the highest exposure taking into
Fabian M Dayrit et al.
Food additives and contaminants, 21(3), 204-209 (2004-06-16)
3-Monochloro-1,2-propane diol is a suspected carcinogen found in hydrolysed vegetable protein products such as soy sauce. A method is described for the analysis of 3-monochloro-1,2-propane diol in soy sauce by gas chromatography-mass spectrometry at a concentration range of 1-5000 ng
M Statheropoulos et al.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences, 822(1-2), 112-117 (2005-07-06)
A preliminary investigation on the possibility of using volatile organic compounds (VOCs) determination of expired air, blood and urine, for the early location of entrapped people in earthquakes, has been carried out. A group of 15 healthy subjects has been
H M Liebich
Journal of chromatography, 273(1), 67-75 (1983-03-11)
Total 4-heptanone is the sum of a beta-oxocarboxylic acid, 2-ethyl-3-oxohexanoic acid, and its decarboxylation product, 4-heptanone. The beta-oxocarboxylic acid is found in serum and in urine, and is detected by gas chromatography--mass spectrometry in the form of its methyl ester

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service