Skip to Content
Merck
All Photos(1)

Key Documents

46149

Supelco

Corticosterone 21-acetate

VETRANAL®, analytical standard

Synonym(s):

11β,21-Dihydroxy-4-pregnene-3,20-dione 21-acetate, 4-Pregnene-11β,21-diol-3,20-dione 21-acetate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C23H32O5
CAS Number:
Molecular Weight:
388.50
Beilstein:
3225312
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

VETRANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

[H][C@@]12CCC3=CC(=O)CC[C@]3(C)[C@@]1([H])[C@@H](O)C[C@]4(C)[C@H](CC[C@@]24[H])C(=O)COC(C)=O

InChI

1S/C23H32O5/c1-13(24)28-12-20(27)18-7-6-17-16-5-4-14-10-15(25)8-9-22(14,2)21(16)19(26)11-23(17,18)3/h10,16-19,21,26H,4-9,11-12H2,1-3H3/t16-,17-,18+,19-,21+,22-,23-/m0/s1

InChI key

WKQCPUMQBMFPLC-ZWFCQKKLSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

VETRANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

L E Flores et al.
Comparative biochemistry and physiology. Part C, Pharmacology, toxicology & endocrinology, 120(1), 77-81 (1998-11-25)
The aim of this work was to demonstrate the possible direct effect of several hormones upon glucose-induced insulin secretion in amphibians. Hence, pancreas pieces of Bufo arenarum were incubated for 60 min at 25 degrees with 2 and 8 mM
Matthew N Hill et al.
European journal of pharmacology, 528(1-3), 99-102 (2005-12-06)
This research was designed to examine the effect of three weeks of administration of corticosterone (20 mg/kg) on endocannabinoid content and cannabinoid CB1 receptor binding in the amygdala. It was found that the endocannabinoid 2-arachidonylglycerol was significantly increased in the
B L Tepperman et al.
The American journal of physiology, 250(5 Pt 1), G617-G624 (1986-05-01)
Gastric mucosal responses to intraluminal instillation of acid (10, 50, and 150 mM HCl) and acidified solutions of sodium taurocholate (10 mM) were measured in rats between 5 and 60 days after birth. Net loss of H+ and gain of
R Chirino et al.
Endocrinology, 129(6), 3118-3124 (1991-12-01)
The low affinity glucocorticoid binding sites (LAGS) have been described and partially characterized in both the nuclei and microsomes of rat liver. The LAGS concentration is under endocrine regulation, as proved by their decrease after adrenalectomy and their almost complete
U D McCann et al.
Endocrinology, 114(5), 1513-1521 (1984-05-01)
Two types of iodothyronine 5'-deiodination have been characterized previously in rat tissues. They can be distinguished by inhibition of type I but not type II by 6-n-propylthiouracil, by the relative suitability of T4 and rT3 as substrates, rT3 much better

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service