Skip to Content
Merck
All Photos(1)

Key Documents

45896

Supelco

Geranyl acetate

analytical standard

Synonym(s):

trans-3,7-Dimethyl-2,6-octadien-1-yl acetate, trans-3,7-Dimethyl-2,6-octadienyl acetate

Sign Into View Organizational & Contract Pricing

Select a Size

1 ML
$63.80

$63.80


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 ML
$63.80

About This Item

Linear Formula:
CH3CO2CH2CH=C(CH3)CH2CH2CH=C(CH3)2
CAS Number:
Molecular Weight:
196.29
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

$63.80


Check Cart for Availability

Request a Bulk Order

grade

analytical standard

Quality Level

vapor density

6.8 (vs air)

vapor pressure

0.07 mmHg ( 20 °C)

Assay

≥99.0% (GC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

refractive index

n20/D 1.461 (lit.)
n20/D 1.462

bp

138 °C/25 mmHg (lit.)

application(s)

agriculture
cleaning products
cosmetics
environmental
flavors and fragrances
food and beverages
personal care

format

neat

storage temp.

2-8°C

SMILES string

CC(=O)OC\C=C(/C)CC\C=C(/C)C

InChI

1S/C12H20O2/c1-10(2)6-5-7-11(3)8-9-14-12(4)13/h6,8H,5,7,9H2,1-4H3/b11-8+

InChI key

HIGQPQRQIQDZMP-DHZHZOJOSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Skin Irrit. 2 - Skin Sens. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

219.2 °F - closed cup

Flash Point(C)

104 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Satoshi Nojima et al.
PloS one, 6(3), e18178-e18178 (2011-04-06)
Semiochemicals are often produced in infinitesimally small quantities, so their isolation requires large amounts of starting material, not only requiring significant effort in sample preparation, but also resulting in a complex mixture of compounds from which the bioactive compound needs
Bio-degradation of acetates of geraniol, nerol & citronellol by P. incognita: isolation & identification of metabolites.
K M Madyastha et al.
Indian journal of biochemistry & biophysics, 20(3), 136-140 (1983-06-01)
Luca Nicolotti et al.
Journal of chromatography. A, 1360, 264-274 (2014-08-19)
Comprehensive two-dimensional gas chromatography (GC×GC) coupled with Mass Spectrometry (MS) is one of today's most powerful analytical platforms for detailed analysis of medium-to-high complexity samples. The column set usually consists of a long, conventional-inner-diameter first dimension ((1)D) (typically 15-30m long
Oriane Tascone et al.
Analytical and bioanalytical chemistry, 406(30), 8041-8048 (2014-10-27)
Damascena and centifolia roses are cultivated worldwide for their petal extracts that contain key odorant ingredients of perfumes. The analytical identification and quantification of pesticides in rose petals have never been described in the literature. Here, we report on a
Pingxi Xu et al.
Proceedings of the National Academy of Sciences of the United States of America, 111(46), 16592-16597 (2014-10-29)
Insect repellents are important prophylactic tools for travelers and populations living in endemic areas of malaria, dengue, encephalitis, and other vector-borne diseases. DEET (N,N-diethyl-3-methylbenzamide) is a 6-decade-old synthetic repellent, which is still considered the gold standard of mosquito repellents. Mosquitoes

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service