Skip to Content
Merck
All Photos(1)

Key Documents

45661

Supelco

Strychnine

PESTANAL®, analytical standard

Synonym(s):

(−)-Strychnine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H22N2O2
CAS Number:
Molecular Weight:
334.41
Beilstein:
52979
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

mp

284-286 °C (lit.)

application(s)

agriculture
cleaning products
cosmetics
environmental
food and beverages
forensics and toxicology
personal care
pharmaceutical (small molecule)

format

neat

SMILES string

[H][C@@]12CC(=O)N3c4ccccc4[C@]56CC[N@H]7CC(=CCO1)[C@]([H])(C[C@@H]57)[C@]2([H])[C@]36[H]

InChI

1S/C21H22N2O2/c24-18-10-16-19-13-9-17-21(6-7-22(17)11-12(13)5-8-25-16)14-3-1-2-4-15(14)23(18)20(19)21/h1-5,13,16-17,19-20H,6-11H2/t13-,16-,17-,19-,20-,21+/m0/s1

InChI key

QMGVPVSNSZLJIA-FVWCLLPLSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Strychnine is an alkaloid, available as a bioactive component in Semen Strychni, exhibiting analgesic, anti-inflammatory and anti-tumor effects. It is widely used in neurological research, treatment of genetic disorder nonketotic hyperglycinemia, treatment of stomach ailments, etc.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.
Strychnine may be used as an analytical reference standard for the quantification of the analyte in biological samples using liquid chromatography coupled to electrospray ionization mass spectrometry.

Biochem/physiol Actions

Convulsant; glycine receptor antagonist not associated with the NMDA receptor.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Skull and crossbonesEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 1 Dermal - Acute Tox. 1 Oral - Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 2

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Application of solid phase microextraction to the determination of strychnine in blood
Borroso.M, et al.
Journal of Chromatography. B, Biomedical Sciences and Applications, 816(1-2), 29-34 (2005)
The 100 Most Important Chemical Compounds: A Reference Guide (2005)
Determination of strychnine and brucine in rat plasma using liquid chromatography electrospray ionization mass spectrometry
Xu Y, et al.
Journal of Pharmaceutical and Biomedical Analysis, 49(2), 487-491 (2009)
Sébastien Dutertre et al.
The Journal of biological chemistry, 287(48), 40216-40223 (2012-10-06)
Strychnine-sensitive glycine receptors (GlyRs) mediate synaptic inhibition in the spinal cord, brainstem, and other regions of the mammalian central nervous system. In this minireview, we summarize our current view of the structure, ligand-binding sites, and chloride channel of these receptors
Yan Lu et al.
The Journal of clinical investigation, 123(9), 4050-4062 (2013-08-28)
Neuropathic pain is characterized by mechanical allodynia induced by low-threshold myelinated Aβ-fiber activation. The original gate theory of pain proposes that inhibitory interneurons in the lamina II of the spinal dorsal horn (DH) act as "gate control" units for preventing

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service