Skip to Content
Merck
All Photos(1)

Documents

45624

Supelco

Dipropyl phthalate

PESTANAL®, analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H4-1,2-(CO2CH2CH2CH3)2
CAS Number:
Molecular Weight:
250.29
Beilstein:
2332522
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

refractive index

n20/D 1.497 (lit.)

bp

317.5 °C (lit.)

density

1.078 g/mL at 25 °C (lit.)

application(s)

agriculture
cleaning products
cosmetics
environmental
food and beverages
personal care

format

neat

SMILES string

CCCOC(=O)c1ccccc1C(=O)OCCC

InChI

1S/C14H18O4/c1-3-9-17-13(15)11-7-5-6-8-12(11)14(16)18-10-4-2/h5-8H,3-4,9-10H2,1-2H3

InChI key

MQHNKCZKNAJROC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Dipropyl phthalate is a low-toxic, broad-spectrum antimicrobial agent belonging to the group of phthalates and is extensively used as a preservative in cosmetic industry.

Application

Dipropyl phthalate may be used as an analytical reference standard for the quantification of the analyte in cosmetic and plastic products using different chromatography techniques.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Health hazardEnvironment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 2 - Repr. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

228.2 °F - closed cup

Flash Point(C)

109 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Simultaneous screening and determination eight phthalates in plastic products for food use by sonication-assisted extraction/GC-MS methods
Shen Y-H
Talanta, 66(3), 734-739 (2005)
Reproductive toxicology. Di-n-propylphthalate.
Environmental health perspectives, 105 Suppl 1, 257-258 (1997-02-01)
Takashi Maruyama et al.
Microbiology and immunology, 51(3), 321-326 (2007-03-24)
Phthalate esters with short alkyl chains, such as di-ethyl (DEP), di-n-propyl (DPP), and di-butyl phthalate (DBP), have adjuvant effects on an FITC-induced contact hypersensitivity mouse model. The adjuvant effects of DPP and DBP are associated with enhanced trafficking of FITC-presenting
Y Imai et al.
Clinical and experimental allergy : journal of the British Society for Allergy and Clinical Immunology, 36(11), 1462-1468 (2006-11-07)
Many different types of phthalate ester are used as plasticizers and are thus found in the air. There have been several studies that suggest an association between allergies and phthalate esters. We previously found that di-butyl phthalate (DBP) has an
Hung-Hung Sung et al.
Fish & shellfish immunology, 31(3), 511-515 (2011-07-12)
This study set out to understand the sublethal effect of xenobiotic phthalate esters (PAEs) on the relationship between the susceptibility of shrimp to bacterial infection and the immune response of the shrimp. Neocaridina denticulate were exposed to different concentrations of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service